Skip to Content
MilliporeSigma
All Photos(2)

Documents

SYMBDSL01

Millipore

Steritest® Symbio drain tray support for 2 canisters, ISL pump

This accessory streamlines the workflow and increases safety during sterility testing

Synonym(s):

Canister holder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151818
NACRES:
NB.79

material

316L stainless steel support (Drain tray)
EPDM rubber seal

Agency

EP (2.6.1)
JP (4.06)
USP (71)

packaging

pkg of 24 units

manufacturer/tradename

Steritest®

input

pharmaceutical(s)

application(s)

pharmaceutical
sterility testing

compatibility

for use with Steritest® Symbio pump

shipped in

ambient

General description

Steritest® Symbio drain tray support includes the stainless steel canister holder and quick connector. This tray is designed for 2 canisters ISL pump.

Application

Accessories for Streamlining Your Workflow and Increasing Safety during sterility testing.

Legal Information

STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service