Key Documents
SYCNA4VP1
Millipore
Steritest® Symbio Pump Validation Protocol
For 3 Canister Pump
About This Item
UNSPSC Code:
41116500
NACRES:
NB.79
Pricing and availability is not currently available.
Quality Level
compatibility
for use with Steritest® Symbio Pumps (For 3 Canister Pumps)
General description
Steritest® Symbio Pump validation protocol is designed to make it easier for the technicians to ensure compliance of the Symbio pumps for consistent performance. The product is GMP compliant and includes Validation Master Plan (VMP), Installation Qualification (IQ), Operational Qualification (OQ) and Performance Qualification (PQ) test protocols. In addition to the above, ready to be filled report table formats are also provided.
Application
Steritest® Symbio Pump validation protocol is a ready to use GMP compliant validation protocol along with ready to be filled report table formats.
Legal Information
STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany