Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ST1650

Sigma-Aldrich

Anti-DYRK1A Mouse mAb (7D10)

liquid, clone 7D10, Calbiochem®

Synonym(s):

Anti-Dual-Specificity Tyrosine-(Y)-Phosphorylation Regulated Kinase 1A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
clone:
7D10, monoclonal
application:
species reactivity:
mouse, human, rat
citations:
1

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

7D10, monoclonal

form

liquid

does not contain

preservative

species reactivity

mouse, human, rat

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG2b

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DYRK1A(1859)
mouse ... Dyrk1A(13548)
rat ... Dyrk1A(25255)

General description

Anti-DYRK1A, mouse monoclonal, clone 7D10, recognizes the ~80-85 kDa DYRK1A in PC-12, 293, HeLa NE, Raw 264.7, & NIH3T3 cells. It is validated for use in ELISA and Western blotting.
Purified mouse monoclonal antibody. Recognizes the ~80-85 kDa DYRK1A protein.
Recognizes the ~80-85 kDa DYRK1A protein in PC-12, 293, HeLa NE, Raw 264.7, and NIH3T3 cells.

Immunogen

A recombinant polypeptide corresponding to amino acids of 674-763 human DYRK1A, expressed as a GST fusion protein

Warning

Toxicity: Regulatory Review (Z)

Analysis Note

Positive Control
PC-12 cells, 293 cells, HeLa NE cells, Raw 264.7 cells, NIH3T3 cells

Other Notes

Guo, X., et al. 2010. J. Biol. Chem.285, 13223.
Guedj, F., et al. 2009. PLoS One.4, e4606.
Hamelet, J., et al. 2009. Biochem. Biophys. Res. Commun.378, 673.
Lee, Y., et al. 2009. J. Biol. Chem.284 33343.
Noll, C., et al. 2009. PLoS One 2009. 4, e7540.
Scales, T.M., et al. 2009. J. Cell Sci.122, 2424.
Aranda, S., et al. 2008. Mol. Cell Biol.28, 5899.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service