Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SLM-020

Sigma-Aldrich

Dulbecco′s Modified Eagle Media 1X, Liquid, w/4,500 mg/L Glucose, L-Glutamine, without Sodium Pyruvate (1X) - 1L

The Dulbecco′s Modified Eagle Media 1X, Liquid, w/4,500 mg/L Glucose, L-Glutamine, without Sodium Pyruvate (1X) is available in a 1 L format & has been optimized & validated for cell culture.

Synonym(s):

DMEM 1X

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
eCl@ss:
32160801
NACRES:
NA.75

Quality Level

form

liquid

manufacturer/tradename

Specialty Media

technique(s)

cell culture | mammalian: suitable

input

sample type hematopoietic stem cell(s)
sample type: mouse embryonic stem cell(s)
sample type: human embryonic stem cell(s)
sample type neural stem cell(s)
sample type mesenchymal stem cell(s)
sample type epithelial cells
sample type cardiac stem cell(s)
sample type pancreatic stem cell(s)
sample type induced pluripotent stem cell(s)

Storage and Stability

Store at 2-8°C for 10 months

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service