Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SLHP033R

Millipore

Millex-HP Filter Unit (Sterile)

pore size 0.45 μm, diam. 33 mm, polyethersulfone membrane, hydrophilic, medical device

Synonym(s):

disposable syringe filter, polyethersulfone syringe filter, sterile syringe filter, syringe filter

Sign Into View Organizational & Contract Pricing

Select a Size

50 PKG
$321.00
250 PKG
$1,240.00

$321.00


Available to ship onApril 07, 2025Details



Select a Size

Change View
50 PKG
$321.00
250 PKG
$1,240.00

About This Item

UNSPSC Code:
41104922
eCl@ss:
32031690
NACRES:
NB.24

$321.00


Available to ship onApril 07, 2025Details


material

modified acrylic housing
polyethersulfone membrane

Quality Level

sterility

sterile; γ-irradiated

product line

Millex

feature

CE compliant
holdup volume≤ 0.1 mL
hydrophilic
medical device

manufacturer/tradename

Millex

parameter

10.3 bar max. inlet pressure (150 psi)
45 °C max. temp.

diam.

33 mm

filtration area

4.52 cm2

inlet to outlet H

27 mm

volume

200 mL

pore size

0.45 μm pore size

fitting

female Luer-Lok® inlet
male Luer outlet slip

shipped in

ambient

General description

Millex-HP Filter Unit (Sterile) is a medical device intended for use as a syringe filter to clarify low volume solutions in direct patient care and pharmacy admixture applications.

Application

  • Clinical benefits: Removal of microorganisms, particles, precipitates, and undissolved powders larger than the membrane’s rated pore size from solutions for clinical use.
  • Hospital pharmacy: Clarification of small volumes of protein pharmaceuticals, diagnostic imaging agents, chemotherapeutics, aqueous solutions, or water during admixture preparation.
  • Direct patient care: Particulate removal from epidural and other liquid anaesthetics as well as from irrigation solutions used in ophthalmic, otic, and other surgical procedures.

Features and Benefits

  • Bidirectional support of the filter membrane
  • Removes microorganisms, particles, precipitates, and undissolved powders larger than the membrane′s rated pore size
  • Single-use, non-pyrogenic and non-toxic
  • Polyethersulfone membrane filter sealed in a modified acrylic copolymer (MMA) housing
  • Compatible with most aqueous solutions
  • CE marked

Other Notes

Devices are intended for medical and/or laboratory professional use only.

Legal Information

Luer-Lok is a registered trademark of Becton-Dickinson & Co.
Millex is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lorane Texari et al.
STAR protocols, 2(1), 100358-100358 (2021-03-16)
Integrative analysis of next-generation sequencing data can help understand disease mechanisms. Specifically, ChIP-seq can illuminate where transcription regulators bind to regulate transcription. A major obstacle to performing this assay on primary cells is the low numbers obtained from tissues. The
Maartje J Levels et al.
Journal of autoimmunity, 101, 131-144 (2019-05-06)
During T cell-dependent (TD) germinal center (GC) responses, naïve B cells are instructed to differentiate towards GC B cells (GCBC), high-affinity long-lived plasma cells (LLPC) or memory B cells (Bmem). Alterations in the B cell-fate choice could contribute to immune
Hongshi Jin et al.
Nucleic acids research, 46(22), 11712-11725 (2018-09-22)
Plant immunity depends on massive expression of pathogenesis-related genes (PRs) whose transcription is de-repressed by pathogen-induced signals. Salicylic acid (SA) acts as a major signaling molecule in plant immunity and systemic acquired resistance triggered by bacterial or viral pathogens. SA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service