SLHA033
Millex™-HA Filter Unit (Sterile)
pore size 0.45 μm, diam. 33 mm, mixed cellulose esters (MCE) membrane, hydrophilic, medical device
Synonym(s):
disposable syringe filter, mixed cellulose esters syringe filter, sterile syringe filter, syringe filter
Select a Size
$315.00
Select a Size
About This Item
$315.00
Recommended Products
material
mixed cellulose esters (MCE) membrane
modified acrylic housing
Quality Level
sterility
sterile; ethylene oxide treated
product line
Millex™
feature
CE compliant
holdup volume≤ 0.1 mL after air purge
hydrophilic
medical device
manufacturer/tradename
Millex™
parameter
150 psi max. inlet pressure
45 °C max. temp.
diam.
33 mm
filtration area
4.52 cm2
inlet to outlet H
27 mm
volume
100 mL
matrix
MF-Millipore™
pore size
0.45 μm pore size
fitting
female Luer-Lok® inlet
male Luer outlet slip
General description
Application
- Clinical benefits: Removal of microorganisms, particles, precipitates, and undissolved powders larger than the membrane’s rated pore size from solutions for clinical use.
- Hospital pharmacy: Clarification of small volumes of protein pharmaceuticals, diagnostic imaging agents, chemotherapeutics, aqueous solutions, or water during admixture preparation.
- Direct patient care: Particulate removal from epidural and other liquid anaesthetics as well as from irrigation solutions used in ophthalmic, otic, and other surgical procedures.
Features and Benefits
- Bidirectional support of the filter membrane
- Removes microorganisms, particles, precipitates, and undissolved powders larger than the membrane′s rated pore size
- Single-use, non-pyrogenic and non-toxic
- Mixed cellulose esters (MCE) membrane filter sealed in a modified acrylic copolymer (MMA) housing
- Compatible with most aqueous solutions
- CE marked
Other Notes
Legal Information
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Sol. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service