Skip to Content
MilliporeSigma
All Photos(1)

Documents

SCM106

Sigma-Aldrich

Human OPC Spontaneous Differentiation Culture Media Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.32

description

Neural Stem Cells

Quality Level

type

for cell culture

form

liquid

technique(s)

cell culture | stem cell: suitable

General description

Human OPC Spontaneous Differentiation Media is a defined, serum-free medium that supports the differentiation of human ESC-derived OPCs (Catalog No. SCR600) to mature oligodendrocytes through growth factor withdrawal. The media does not support the differentiation of primary oligodendrocyte progenitor cells isolated from brain tissues

Storage and Stability

Store at -20°C, good for 4 months from date of receipt when reagents are handled and stored appropriately. Please note: The supplemented Human OPC Spontaneous Differentiation cOmplete Media should be stored at 2 to 8C for up to 4 weeks.

Legal Information

cOmplete is a trademark of Roche

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service