Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SCR600

Sigma-Aldrich

Human Oligodendrocyte Differentiation Kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$1,610.00

$1,610.00


Please contact Customer Service for Availability


Select a Size

Change View
1 KIT
$1,610.00

About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.32

$1,610.00


Please contact Customer Service for Availability

description

Cell line type: Neural Stem Cells
Neural Stem Cells

Quality Level

technique(s)

cell culture | stem cell: suitable

Components

Human Oligodendrocyte Progenitors (OPCs), 500,000 viable cells: (Cat. No. CS204496) Human OPC Expansion Media Kit, 100 mLs: (Cat. No. SCM107) Human OPC Spontaneous Differentiation Media Kit, 100 mLs: (Cat. No. SCM106)

Quality

Sterility testing:Negative for bacterial and fungal growth
pH:7.4 +/- 0.4
Osmolality:290 +/- 20mOsm

Storage and Stability

Please see manual for recommended storage conditions for individual components

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service