Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SCC245

YUMMER1.7-H2B-GFP5 Mouse Melanoma Cell Line

The YUMMER1.7-H2B-GFP5 mouse melanoma cell line is a valuable model for studies of immune checkpoint inhibition and mechanisms of anti-tumor responses.

Synonym(s):

Melanoma Cell Culture

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

application(s)

cell analysis

General description

The YUMMER1.7 H2B-GFP mouse melanoma cell line is both immunocompetent and reflective of the somatic mutations common in melanomas. The YUMMER1.7 H2B-GFP cell line carries three driver mutations of melanoma: Braf V600E, Pten -/- and Cdkn2 -/- (1,2). In addition, the YUMMER1.7 H2B-GFP cell line harbors a high frequency of stable UV-induced somatic mutations which have been shown to stimulate host adaptive immune response (3). YUMMER1.7-H2B-GFP cells stably express GFP fused to histone H2B, allowing visualization of chromosomal dynamics and facilitating in vivo detection of YUMMER1.7 melanoma cells and YUMMER1.7-derived tumors. The unique features of the YUMMER1.7 H2B-GFP cell line make it a valuable model for studies of immune checkpoint inhibition and mechanisms of anti-tumor responses.

<bold>Source:</bold>
The YUMMER1.7 H2B-GFP mouse melanoma cell line is a tetraploid clonal isolate derived from YUMM1.7 cells exposed to UVB radiation into which a histone H2B-GFP fusion has been stably introduced via transfection. The original YUMM1.7 cell line was derived from a 4-hydroxytamoxifen-induced melanoma tumor in a male C57/B1/6 mouse into which mutations from the Braf/Pten genetically-engineered mouse model had been introduced via backcrossing (1). The YUMMER1.7 H2B-GFP cell line harbors the Braf V600E mutation and is homozygous negative for wild-type Pten and Cdkn2 (3).

Research Category:
Cancer

Cell Line Origin

Mouse, Cancer Cells

Packaging

&ge;1X106 cells/vial

Storage and Stability

Store in liquid nitrogen. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.

Other Notes

This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the “Academic Use Agreement” as detailed in the product documentation. For information regarding any other use, please contact [email protected].

Disclaimer

This product contains genetically modified organisms (GMO).
Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service