Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SCC265

Sigma-Aldrich

Huh-7.5 Tet-On Human Hepatocellular Carcinoma Cell Line

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.45

biological source

human

packaging

vial of >1X10⁶ cells

manufacturer/tradename

Millipore

growth mode

N/A

technique(s)

cell culture | mammalian: suitable

shipped in

liquid nitrogen

storage temp.

−196°C

Application

  • Each vial contains >1X106 viable cells.
  • Cells are tested negative for infectious diseases by a Human Essential CLEAR panel by Charles River Animal Diagnostic Services.
  • Cells are verified to be of human origin and negative for inter-species contamination from rat, mouse, chinese hamster, Golden Syrian hamster, and non-human primate (NHP) as assessed by a Contamination CLEAR panel by Charles River Animal Diagnostic Services.
  • Cells are negative for mycoplasma contamination.
  • Each lot of cells is genotyped by STR analysis to verify the unique identity of the cell line.

Huh-7 is a well differentiated hepatocyte-derived carcinoma cell line, isolated from the liver tumor of a 57-year-old Japanese male (Nakabayashi, 1982). Derived from Huh-7, the Huh-7.5 cell line is highly permissive for subgenomic and genomic Hepatitis C virus (HCV) RNA replication (Blight et al., 2002) and is now employed for high yield production of HCV and studies of antivirals on industrial scale. Huh-7.5 Tet-On Human Hepatocellular Carcinoma Cell Line was generated from Huh-7.5 cells using retroviral transduction to express the Tet-On 3G transactivator protein and enable subsequent doxycycline-regulated transgene expression to aid in the study of subcellular interactions between HCV and its host (Luna, 2015; Takacs, 2017).

Features and Benefits

Huh-7.5 Tet-On human hepatocellular carcinoma cell line is useful for Hepatitis C virus (HCV) studies and production.

Storage and Stability

Cells should be stored in liquid nitrogen until use. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.

Other Notes

This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the “Academic Use Agreement” as detailed in the product documentation.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service