Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SCC132M

Sigma-Aldrich

CCMCL1 Mantle Cell Lymphoma Cell Line

Human

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.81
Pricing and availability is not currently available.

Product Name

CCMCL1 Mantle Cell Lymphoma Cell Line, CCMCL1 human mantle cell lymphoma cell line is well characterized at the molecular level and can support research and drug development efforts targeting MCL.

biological source

human

Quality Level

technique(s)

cell culture | mammalian: suitable

General description

Mantle cell lymphoma (MCL) is incurable using current standard therapeutic regimens. CCMCL1 cell line is a new cell culture model of aggressive mantle cell lymphoma (1). Primary MCL cells of a 58 year old man with progressive MCL and hyperleukocytosis were engrafted in NOD-SCID-gamma mice. Splenocytes obtained from the engrafted mouse were cultured in vitro and yielded the CCMCL1 cell line. CCMCL1 cells possess a blastoid nuclear morphology and proliferate readily in suspension culture. CCMCL1 cells express CD19, CD20, and CD45 and are negative for CD3 and CD25.

CCMCL1 has the IGH-CCND1 genetic hallmark of MCL and closely resembles the terminal-phase leukemic MCL cells as evidenced by immunophenotyping, karyotyping, IGHV sequence, and whole exome sequencing (WES) (1). WES provides unbiased, direct evidence for conservation of the MCL genome in CCMCL1 at single-nucleotide resolution. WES detected no mutation in genes of the B-cell receptor or phosphatidylinositol 3-kinase pathway, including BTK and AKT. However, CCMCL1 constitutively expressed phosphorylated AKT and BTK, suggesting CCMCL1 is an appropriate model for studies targeting these two pathways in MCL. Other MCL cell lines have mutations or deletions in either ATM or TP53; with this respect CCMCL1 also offers an opportunity for MCL studies without the interference of ATM or TP53 loss.

Reference:
1. Zhao X, Chen-Kiang S, Shetty S, Di Liberto M, Bodo J, Durkin L, Eng K, Elemento O, Smith MR, Hsi ED (2015) CCMCL1: a new model of aggressive mantle cell lymphoma. Blood 125(17): 2730-2732.

Cell Line Description

Cancer Cells

Application

CCMCL1 human mantle cell lymphoma cell line is well characterized at the molecular level and can support research and drug development efforts targeting MCL.
Research Category
Cancer

Oncology
This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the “Academic Use Agreement” as detailed in the product documentation. For information regarding any other use, please contact [email protected].

Quality

• Each vial contains ≥ 1X106 viable cells.
• Cells are tested negative for Epstein-Barr virus, HPV-16, HPV-18, Hepatitis A, C, Herpesvirus type 6, 7, 8 and HIV-1 & 2 viruses by PCR
• Cells are negative for mycoplasma contamination.
• Each lot of cells is genotyped by STR analysis to verify the unique identity of the cell line

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service