Skip to Content
MilliporeSigma
All Photos(1)

Documents

MABF2816

Sigma-Aldrich

Anti-Cortical stromal cells Antibody, clone ER-TR4

Synonym(s):

cTEC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

ER-TR4, monoclonal

purified by

affinity chromatography

species reactivity

mouse

packaging

antibody small pack of 100 μg

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable

isotype

IgMκ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

storage temp.

2-8°C

target post-translational modification

unmodified

General description

The thymus is considered as the primary lymphoid organ that is responsible for the generation and maturation of T cells. Thymic epithelial cells (TECs) account for the majority of thymic stromal components. These cells can be divided into cortical and medullary thymic epithelial cells based on their localization within the thymus. TECs have dual germ layer origins, with cortical cells arising from ectoderm and medullary cells arising from the endoderm. In an embryonic thymus, cortical cells are the dominant population, while medullary cells account for most of the TECs in the adult thymus. Cortical thymic epithelial cells are essential for the positive selection of T cells, whereas the medullary thymic epithelial cells (mTECS) play a role in inducing negative selection of highly reactive T cells that are required for establishing central self-tolerance. mTECs participate in self-tolerance by eliminating self-reactive T cells. Cortical cells that support positive selection also support post-selection maturation of single-positive thymocytes. They provide cytokines, such as IL-7 and SCF, to promote early thymic progenitors (ETP) cells and Delta-like ligand 4 (DLL4)-mediated Notch signaling to induce the differentiation of ETPs towards the T lineage. DLL4 is reported to be expressed at high levels in cortical cells, but not in medullary cells. Clone ER-TR4 reacts with epithelial-reticular cells in the cortex of murine thymus. (Ref.: Wang, HX., et al. (2020). Front. Immunol. 10; 3099; Van Vliet, E., et al. (1984). Eur. J. Immunol. 14(6); 524-529; Cordier, AC., et al. (1980). Am. J Anat. 157(3); 227-263).

Specificity

Clone ER-TR4 is a rat monoclonal antibody that detects cortical stromal cells.

Immunogen

Thymic stromal cells isolated from C3H mice.

Application

Quality Control Testing

Evaluated by Immunohistochemistry in mouse thymus tissue sections.

Immunohistochemistry Applications: A 1:50 dilution of this antibody detected Cortical stromal cells in mouse thymus tissue sections.

Tested applications

Immunofluorescence Analysis: A representative lot detected Cortical stromal cells in Immunofluorescence applications (Tanaka, Y., et al., (2007). Blood. 110(1):107-15).

Immunohistochemistry Applications: A representative lot detected Cortical stromal cells in Immunohistochemistry applications (Van Vliet, E., et al., (1984). Eur J Immunol. 14(6):524-9; Scheijen, B., et al., (2004). J Biol Chem. 279(11):10476-83; Dooley, J., et al., (2006). J Immunol. 176(11):6484-90; Tanaka, Y., et al., (2007). Blood. 110(1):107-15; Moretti, F.A., et al., (2018). Elife. 7:e35816; Hollander, G.A., et al. (1995). Nature. 373(6512):350-3).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Physical form

Purified rat monoclonal antibody IgM in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Reconstitution

1.0 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service