Key Documents
RDHSPACE1
Milli-Q
Spacer for Remote Dispenser
For use with Direct-Q® and Synergy® Water Purification Systems
Select a Size
1 EA
$310.00
$310.00
Please contact Customer Service for Availability
Request more information
Select a Size
Change View
1 EA
$310.00
About This Item
UNSPSC Code:
41104200
NACRES:
SB.51
$310.00
Please contact Customer Service for Availability
Request more information
packaging
pkg of 1 unit
compatibility
for use with Direct-Q® UV
for use with Direct-Q®
for use with Synergy® UV
for use with Synergy®
General description
The spacer is used when the installation requires the dispenser to be wall-mounted. This accessory increases the distance between the dispenser outlet and the wall. This additional space is required for the dispenser to be positioned right above a sink. It can also be useful when placing large glassware conveniently under the dispenser outlet.
Application
Dispensers mounted on a wall can be used with a spacer to gain more depth.
Legal Information
DIRECT-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
SYNERGY is a registered trademark of Merck KGaA, Darmstadt, Germany