Documents
Z563064
Work station anti-static ionizer for Plas-Labs glove box
AC/DC input 120 V AC
Synonym(s):
humidity chambers, humidity control chambers, isolation chambers, nitrogen dry box
About This Item
UNSPSC Code:
41103406
NACRES:
NB.35
material
electrode (Thoriated tungsten)
manufacturer/tradename
Plas-Labs 800-AS/SPI
availability
not available in EU
Overall H × W × depth
5.0 in. × 4 1/4 in. × 2 1/2 in.
weight
2 lb
General description
The anti-static ionizer effectively eliminates all static charges within 36 inches of the unit and is useful in any Plas-Labs glove box or isolation chamber.
Operation is simple; place the unit on a flat surface and turn the power on.
Unit features include: ion pulse rate adjustment from 1 to 5 pulses/second, ion balance adjustment and a compact footprint. The unit is virtually maintenance free; the emitter points should be cleaned when a visual accumulation of dust or powder occurs.