Documents
PA51/111
Millipore
Pneumatic Plastic Actuator for NOVASEPTIC® Valve, Normally Closed (NC)
About This Item
UNSPSC Code:
31251504
material
plastic cover (POM C)
plastic indicator (POM C)
stainless steel cover
Quality Level
Agency
meets EN ASTM 303
meets EN Stainless Steel EN 1.4305
parameter
-6-(-1) bar pressure range
0-70 °C temp. range
roughness average (RA)
1.6 μm ((63 μin) Electropolished)
weight
4.4 kg (9.70 lb)
Application
The applied valve body and diaphragm may have different design temperature and/or pressure limits. The weakest component in the assembled product determines the maximum design temperature and pressure limits.
Packaging
The Actuator is packaged in a closed box.
Legal Information
NOVASEPTIC is a registered trademark of Merck KGaA, Darmstadt, Germany