P2C005C05HC
Pellicon® 2 Cassette with Ultracel® 5 kDa Membrane, C screen, 0.5 m²
Select a Size
$6,732.00
Select a Size
About This Item
$6,732.00
Recommended Products
material
composite regenerated cellulose (CRC)
polypropylene
polypropylene screen
urethane foam-filled high-density polyethylene
Quality Level
description
Filter code PLCCC
Type C screen (coarse screen)
prefiltration requirements: 100 µm
parameter
0-30 °C max. temp.
0.3 bar max. transmembrane pressure (5 psig) at 30 °C (Reverse)
3.4 bar max. transmembrane pressure (50 psig) at 30 °C (Forward)
4-7 L/min-cm2 recirculation rate
technique(s)
analytical sample preparation: suitable
protein purification: suitable
tangential flow filtration (TFF): suitable
ultrafiltration: suitable
L
20.6 cm (8.1 in.)
membrane nominal area
0.1 m2
width
17.8 cm , 7.0 in.
matrix
Ultracel®
pore size
5 kDa filter
pH range
2-13
Analysis Note
Legal Information
Disclaimer
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service