Skip to Content
MilliporeSigma
All Photos(1)

Documents

MDA-MB-231 Cell Line human

92020424, human breast, Epithelial

Synonym(s):

MD Anderson-Metastatic Breast-231 Cells, MDA-MB231 Cells, MDA231-BRE Cells, MDAMB231 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32190102

product name

MDA-MB-231 Cell Line human, human breast adenocarcinoma, 92020424

biological source

human breast

growth mode

Adherent

karyotype

Modal no.′s 62 and 64, near triploid

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian breast adenocarcinoma

Cell Line Description

Isolated from pleural effusions of a breast cancer patient.

Application

MDA-MB-231 has been used to study:
  • the changes in the morphological pattern and gene expression of cells when grown in the presence of sera positive for anti-carbonic anhydrase I (CA I) autoantibodies
  • the effects of normothermic conditioned microwave irradiation on cultured cells using an irradiation system
  • the transendothelial migration rates of the cells under different shear stress levels

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 12,13
D13S317: 13
D16S539: 12
D5S818: 12
D7S820: 8,9
THO1: 7,9.3
TPOX: 8,9
vWA: 15,18

Culture Medium

L15+ 2mM Glutamine + 15% Foetal Bovine SerumFBS / FCS.

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6, i.e. seeding at 3-6x10,000 cells/cm2 using 0.05% Trypsin or Trypsin/EDTA; 37°C. CO2 is not required.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

DNA, RNA, cDNA derived from ECACC mammalian cell lines allow screening for genes or expression patterns to identify lines most suitable for specific research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service