Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

MZAABG101

Millipore

Microfil® with EZ-Pak®

sample volume 100 mL, mixed cellulose esters (MCE), pore size 0.8 μm, filter diam. 47 mm, black filter, gridded filter

Synonym(s):

Membrane filtration system

Sign Into View Organizational & Contract Pricing

Select a Size

150 SETS
$505.00

$505.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
150 SETS
$505.00

About This Item

UNSPSC Code:
41104914
eCl@ss:
32031623
NACRES:
NB.24

$505.00


Please contact Customer Service for Availability

Request more information

material

black filter
gridded filter
mixed cellulose esters (MCE)
polypropylene

Agency

ISO 7704

description

Yeast or mold

sterility

sterile; ethylene oxide treated

feature

easily wetted with water
hydrophilic

manufacturer/tradename

Microfil® with EZ-Pak®

parameter

100 mL sample volume

capacity

100 mL

filter diam.

47 mm

funnel capacity

100 mL

matrix

MF-Millipore

pore size

0.8 μm pore size

input

sample type cosmetics
sample type cider
sample type food(s)
sample(s) (environmental)
sample type sport drink
sample type beverage(s)
sample type wine
sample type beer
sample type soft drink
sample type bottled water

application(s)

bioburden testing
clinical
cosmetics
environmental
environmental
food and beverages
industrial qc

shipped in

ambient

General description

For the ultimate in convenience and efficiency, combine the EZ-Pak system with the Microfil filtration system. The Microfil system includes ready-to-use, sterile filtration funnels and a unique filtration support to facilitate sample processing. No preparation steps are required.
The Microfil filtration system is a simple and reliable solution for routine microbiological testing. These ready-to-use, 100 mL Microfil funnels that comes with the 0.8 μm pore size, black gridded EZ pak filter system eliminate the need for washing and sterilizing after each test thus making the equipment handling easier and efficient to retain and recover microorganism like yeast and molds. The push-fit design of the Microfil funnels seals tightly to manifold supports without the need for clamps or O-rings. Unlike with conventional filter holders, incomplete sealing at the membrane/funnel interface is eliminated resulting in consistent recovery of organisms.

Application

These filters find application in Clinical, Cosmetics, Environmental, Food and Beverage, Industrial QC and Pharmaceutical industry. Microbial analysis of beer, bottled water, cider, sport drink and wine by membrane filtration method.
Yeast or mold

Principle

Vacuum

Physical form

Color Code: Black

Legal Information

EZ-PAK is a registered trademark of Merck KGaA, Darmstadt, Germany
MF-Millipore is a trademark of Merck KGaA, Darmstadt, Germany
MICROFIL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Microfil® filtration system ensures microbiological quality assurance in routine testing of raw materials.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service