Skip to Content
MilliporeSigma
All Photos(1)

Documents

MAGNARIP01

Sigma-Aldrich

MAGNA RIP® Kit, 12 Reactions, 2-8°C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105331
NACRES:
NA.84

Legal Information

MAGNA RIP is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service