Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

17-10520

Sigma-Aldrich

Magna NuCLEAR RIP (Cross-Linked) Nuclear RNA-Binding Protein Immunoprecipitation Kit

Sign Into View Organizational & Contract Pricing

Select a Size

24 ASSAYS
$1,030.00

$1,030.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
24 ASSAYS
$1,030.00

About This Item

UNSPSC Code:
41105331
NACRES:
NA.84

$1,030.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

General description

Magna Nuclear RIP Kits are specially designed to allow the discovery and analysis of a variety of chromatin associated RNAs such as long non-coding RNAs, enhancer RNAs and miRNAs . These chromatin-associated RNAs often regulate gene expression and can be analyzed with applications including quantitative reverse transcription polymerase chain reaction (RT-PCR), microarray analysis (RIP-chip) and next generation sequencing (RIP-Seq).

Nuclear RIP can be performed using chromatin that has interactions stabilized by formaldehyde treatment (cross-linked) or chromatin that has not been treated with a cross linking reagent (native). While both of these approaches are similar in that they are designed to recover chromatin associated RNA, the reagents used and the details of the protocol and types of interactions typically detected are different. Cross-linked can capture higher molecular weight complexes in in vivo configurations with possibly lower affinities. In contrast native RIP is expected to recover high affinity, more direct interactions between proteins encoded RNA binding motifs and candidate RNAs. For less well understood proteins and protein complexes often both approaches are used.

Application

Magna Nuclear RIP (Cross-Linked) RNA-Binding Protein Immunoprecipitation Kit is designed for the analysis of chromatin associated RNA such lncRNAs, enhancer RNAs and miRNAs.

Packaging

Kit capacity: 24 RNA-binding protein immunoprecipitation assays using a cross-linked nuclear lysate

Components

10X Glycine;10X PBS;Nuclei Isolation Buffer;RIP Cross-Linked Lysis Buffer;Protein A/G Magnetic Beads;Nuclear RIP Dilution Buffer;Low Salt Wash Buffer;High Salt Wash Buffer;LiCl Wash Buffer;TE Buffer;RIP Elution Buffer;10% SDS;0.5 M EDTA;DNase I (RNase Free);DNase I Supplement;DNase I Reaction Buffer ;Protease Inhibitor Cocktail III, Animal Free ;RNAse Inhibitor;Proteinase K

Legal Information

NuCLEAR is a trademark of Sigma-Aldrich Co. LLC

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service