Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABS1247

Sigma-Aldrich

Anti-BIG1 Antibody, clone 102

clone 102, from mouse

Synonym(s):

Brefeldin A-inhibited guanine nucleotide-exchange protein 1, Brefeldin A-inhibited GEP 1, ADP-ribosylation factor guanine nucleotide-exchange factor 1, p200 ARF guanine nucleotide exchange factor, p200 ARF-GEP1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$481.00

$481.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$481.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$481.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

102, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... ARFGEF1(10565)

General description

Brefeldin A-inhibited guanine nucleotide-exchange protein 1 (UniProt: Q9Y6D6; also known as BIG1, Brefeldin A-inhibited GEP 1, ADP-ribosylation factor guanine nucleotide-exchange factor 1, p200 ARF guanine nucleotide exchange factor, p200 ARF-GEP1) is encoded by the ARFGEF1 (also known as ARFGEP1, BIG1) gene (Gene ID: 10565) in human. BIG1 is expressed in placenta, lung, heart, brain, kidney and pancreas. BIG1 activates ADP-ribosylation factor (ARF) GTPases and is involved in vesicular trafficking. It promotes the activation of ARF1/ARF3 through replacement of GDP with GTP. BIG1 is required for the maintenance of Golgi structure and this function may be independent of its GEF activity. BIG1 and BIG2 display about 74% homology in their amino acid sequence and about 90% identity in their Sec7 domains. BIG1 is phosphorylated by PKA that reduces its GEF activity. It is localized in the cytoplasm in a multiprotein complex along with BIG2. Its nuclear localization has also been reported and it contains a nuclear localization sequence (aa 711-715). In serum starved cells treated with FK506, BIG1 is essentially completely concentrated in nuclei. BIG1 is shown to be a binding partner of myosin IXb and regulates its Rho-GTPase activating protein activity. Here BIG1 and RhoA compete with each other for the binding to myosin IXb, thus resulting in the inhibition of the GAP activity by BIG1. (Ref.: Lin, S et al. (2013). Arterioscler Thromb Vasc Biol. 33(2): e31-e38).

Specificity

Clone 102 specifically detects BIG1 protein in human and targets an epitope within amino acids 212-522 in the N-terminal region.

Immunogen

GST-tagged recombinat fragment corresponding to 311 amino acids from human Brefeldin A-inhibited guanine nucleotide-exchange protein 1 (BIG1).

Application

Detect BIG1 using this mouse monoclonal Anti-BIG1 Antibody, clone 102, Cat. No. MABS1247. It has been tested in Immunocytochemistry and Western Blotting.
Research Category
Signaling
Western Blotting Analysis: 0.5 µg/mL of this antibody detected BIG1 in 10 µg of A431 and HEK293 cell lysates.

Immunocytochemistry Analysis: A representative lot detected BIG1 in Immunocytochemistry applications (Lowery, J., et. al. (2013). J Biol Chem. 288(16):11532-45).

Western Blotting Analysis: A representative lot detected BIG1 in Western Blotting applications (Lowery, J., et. al. (2013). J Biol Chem. 288(16):11532-45).

Quality

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected BIG1 in 10 µg of Jurkat cell lysate.

Target description

~209 kDa observed; 208.77 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jay M Bhatt et al.
Molecular biology of the cell, 30(12), 1523-1535 (2019-04-04)
The integrity of the Golgi and trans-Golgi network (TGN) is disrupted by brefeldin A (BFA), which inhibits the Golgi-localized BFA-sensitive factor (GBF1) and brefeldin A-inhibited guanine nucleotide-exchange factors (BIG1 and BIG2). Using a cellular replacement assay to assess GBF1 functionality

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service