Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.46729

Millipore

Malt Extract Agar with Chloramphenicol

for yeasts, for molds, pkg of (90 mm plate (20 plates per box))

Synonym(s):

Malt Extract Agar with antibiotic

Sign Into View Organizational & Contract Pricing

Select a Size

20 EA
$80.90

$80.90


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
20 EA
$80.90

About This Item

UNSPSC Code:
41106204
NACRES:
NA.74

$80.90


Please contact Customer Service for Availability

Request more information

sterility

sterile; autoclaved

Quality Level

form

plate (ready-to-use)

feature

ready-to-use

packaging

pkg of (90 mm plate (20 plates per box))

pH

5.6 ( in H2O)

application(s)

bioburden testing
food and beverages

storage temp.

2-8°C

suitability

molds
yeasts

General description

Malt Extract Agar with Chloramphenicol is a selective medium for the cultivation, detection, isolation, and determination of yeasts and molds while inhibiting Gram-positive and Gram-negative bacteria. The low pH of the medium promotes the growth of most yeasts and fungi while it inhibits bacterial growth. Malt extract has a high content of carbohydrates (90-92%), along with nitrogen compounds and vitamins (e.g. riboflavin, pyridoxine, and thiamine). The soymeal peptone content (also rich in carbohydrates) additionally supplies the necessary protein compounds such as peptides and amino acids. This composition facilitates the good growth of yeasts, fungi, and dermatophytes. The addition of Chloramphenicol allows a more effective inhibition of bacterial growth while it is less inhibitory to sub-lethally damaged Yeasts and Molds.

Application

Malt Extract Agar with Chloramphenicol is suitable for the enumeration of Yeasts and Molds in food products.

Features and Benefits

Our ready to use media provide the highest level of quality and testing confidence. They have been formulated and tested to meet the pharmacopoeia requirements. Sterility testing media and rinse solutions are manufactured in an ISO 9001, environmentally controlled production center. Each lot undergoes a stringent quality control (QC) procedure, including pH, sterility and growth promotion tests. Our manufacturing approach ensures the highest level of clarity for our media and rinsing fluids, therefore improving accuracy and significantly reducing the risk of incorrect interpretation and false results.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service