Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABN20

Sigma-Aldrich

Anti-HCN1 Antibody, clone N70/28

clone N70/28, from mouse

Synonym(s):

hyperpolarization activated cyclic nucleotide-gated potassium channel 1, brain cyclic nucleotide gated channel 1, Brain cyclic nucleotide-gated channel 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

N70/28, monoclonal

species reactivity

rat

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... HCN1(348980)

General description

HCN1 is a member of a family of pacemaker channels activated by hyperpolarization and regulated by cyclic nucleotides. HCN1 and HCN2 proteins play an important role for motor learning and synapse formation through integration of cerebellar Purkinje cells. The two proteins also shape autonomous activity of single neurons and the periodicity of network oscillations. HCN1 expression is particularly high in the cerebral cortex, hippocampus, cerebellum, and facial motor nucleus. Hyperpolarization of activated cation channels of the HCN gene family such as HCN1, contributes to spontaneous rhythmic activity in both heart and brain. HCN channels pass both Na+ and K+ with relative permeability ratio of ~0.15 to 0.25. The primary sequence of HCN pore region is clearly related to that of highly selective K+ channels. In contrast to nonselective CNG or Na+-channels, both K+ and HCN channels contain a GYG motif in the pore.

Specificity

This antibody recognizes HCN1 at the cytoplasmic domain.

Immunogen

Epitope: Cytoplasmic domain
Recombinant protein corresponding to rat HCN1 at the cytoplasmic domain.

Application

Detect HCN1 using this Anti-HCN1 Antibody, clone N70/28 validated for use in WB, IH.
Immunohistochemistry Analysis: 1:300 dilution from a previous lot detected HCN1 in rat cerebellum and rat brain/inferior colliculus tissue.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters

Quality

Evaluated by Western Blot in rat brain lysate.

Western Blot Analysis: 2 µg/mL of this antibody detected HCN1 on 10 µg of rat brain lysate.

Target description

~120 kDa observed; consistent with a glycosylation of a 100kDa protein.

Physical form

Format: Purified
Protein G
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Rat brain lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Subcellular localization and expression of bamboo mosaic virus satellite RNA-encoded protein.
Palani, PV; Chiu, M; Chen, W; Wang, CC; Lin, CC; Hsu, CC; Cheng, CP; Chen, CM; Hsu, YH; Lin, NS
The Journal of General Virology null
Anikó Rátkai et al.
Scientific reports, 11(1), 3236-3236 (2021-02-07)
Homeostatic plasticity stabilizes neuronal networks by adjusting the responsiveness of neurons according to their global activity and the intensity of the synaptic inputs. We investigated the homeostatic regulation of hyperpolarization-activated cyclic nucleotide-gated (HCN) and T-type calcium (CaV3) channels in dissociated
Joon Ha Park et al.
International journal of molecular medicine, 44(5), 1801-1810 (2019-10-02)
Hyperpolarization‑activated cyclic nucleotide‑gated (HCN) channels have been known to participate in the regulation of neuronal excitability, synaptic transmission and long‑term potentiation in the hippocampus. The present study investigated transient ischemia‑induced changes of HCN1 and HCN2 expressions in the Cornu Ammonis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service