Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB5378

Sigma-Aldrich

Anti-HCN2 Antibody

Chemicon®, from rabbit

Synonym(s):

Hyperpolarization-Activated Cyclic Nucleotide-Gated Potassium Channel 2, BCNG2, HAC-1

Sign Into View Organizational & Contract Pricing

Select a Size

50 μL
$519.00
200 μL
$859.00

$519.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 μL
$519.00
200 μL
$859.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$519.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

Specificity

Recognizes HCN2 protein. The epitope is specific for HCN2 and is not present in any other known protein.

Immunogen

Purified peptide from human HCN2 (amino acids 147-161) (Accession CAB42602).

Application

Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters
This Anti-HCN2 Antibody is validated for use in IH, WB for the detection of HCN2.
Western blot: 1:200-1:1,000 using ECL on rat brain membranes.

Immunohistochemistry on rat brain sections.

Dilutions should be made using a carrier protein such as BSA (1-3%)

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA, 5% sucrose as a stabilizer and 0.05% sodium azide as a preservative. Reconstitute with 50 μL of sterile deionized water. Centrifuge antibody preparation before use (10,000 xg for 5 min).

Storage and Stability

Maintain lyophilized material at -20°C for up to 12 months. After reconstitution maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
CONTROL ANTIGEN:

Included free of charge with the antibody is 40 μg of control antigen (lyophilized powder). The stock solution of the antigen can be made up using 100 μL of sterile deionized water. For negative control, preincubate 1 μg of peptide with 1 μg of antibody for one hour at room temperature. Optimal concentrations must be determined by the end user.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Choong-Hyun Lee et al.
Iranian journal of basic medical sciences, 22(11), 1308-1313 (2020-03-05)
Hyperpolarization-activated cyclic nucleotide-gated (HCN) channels play essential roles in various hippocampal functions, including regulation of long-term potentiation, synaptic plasticity, and hippocampal-dependent cognitive process. The objective of this study was to investigate age-related changes in HCN1 and HCN2 protein expressions in
Amy L Brewster et al.
Neurobiology of disease, 19(1-2), 200-207 (2005-04-20)
Hyperpolarization-activated cyclic nucleotide-gated (HCN) channels mediate hyperpolarization-activated currents (I(h)). In hippocampus, these currents contribute greatly to intrinsic cellular properties and synchronized neuronal activity. The kinetic and gating properties of HCN-mediated currents are largely determined by the type of subunits--for example
Regulation of the hyperpolarization-activated cationic current Ih in mouse hippocampal pyramidal neurones by vitronectin, a component of extracellular matrix
Vasilyev, Dmitry V and Barish, Michael E
The Journal of Physiology, 560, 659-675 (2004)
Xiao-Meng Hao et al.
Frontiers in aging neuroscience, 11, 190-190 (2019-08-14)
The globus pallidus occupies a critical position in the indirect pathway of the basal ganglia motor control system. Hyperpolarization-activated cyclic-nucleotide gated (HCN) channels play an important role in the modulation of neuronal excitability. In vivo extracellular single unit recording, behavioral
Dmitry V Vasilyev et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 22(20), 8992-9004 (2002-10-22)
The hyperpolarization-activated excitatory current I(h) shapes rhythmic firing and other components of excitability in differentiating neurons, and may thus influence activity-dependent CNS development. We therefore studied developmental changes in I(h) and underlying hyperpolarization-activated cyclic nucleotide-gated (HCN) channel subunits in pyramidal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service