Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABF985

Sigma-Aldrich

Anti-Eosinophil-Derived Neurotoxin Antibody, clone 6D1.5/A5

clone 6D1.5/A5, from mouse

Synonym(s):

Non-secretory ribonuclease, Eosinophil-derived neurotoxin, Ribonuclease 2, Ribonuclease US, RNase 2, RNase UpI-2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$622.00

$622.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$622.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$622.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6D1.5/A5, monoclonal

species reactivity

human

technique(s)

ELISA: suitable
multiplexing: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... RNASE2(6036)

General description

Non-secretory ribonuclease (EC 3.1.27.5; UniProt P10153; also known as Eosinophil-derived neurotoxin, Ribonuclease 2, Ribonuclease US, RNase 2, RNase UpI-2) is encoded by the RNASE2 (also known as EDN, RAF3, RNS2) gene (Gene ID 6036) in human. The major basic protein (MBP), eosinophil cationic protein (ECP), eosinophil derived neurotoxin (EDN) and eosinophil peroxidase (EPO), are eosinophil effector molecules implicated in host defense, immunoregulatory responses, and in allergic and inflammatory reactions. These highly cationic proteins are stored preformed within the eosinophil secondary granules and released in intact membrane-bound granules during cell lysis or through a complex system of vesiculotubular structures during “piecemeal necrosis”. While eosinophil count and eosinophil surface expression of the activation marker CD69 are significantly correlated with serum concentrations of MBP, EDN and EPO, studies show that they are not correlated with ECP level. On the other hand, eosinophilic disease patients with normal eosinophil count show significantly increased concentrations of MBP, ECP, EDN, and EPO compared to normal donors. The measurement of MBP, ECP, EDN, and EPO levels thus provides a better predictor than eosinophil count alone in disease diagnosis. EDN is produced with a signal peptide sequence (a.a. 1-27), the removal of which yields the mature (a.a. 28-161) protein.

Specificity

Clone 6D1.5/A5 specifically reacted with EDN, but not other granule proteins by Western blotting analysis (Makiya, M.A., et al. (2014). J. Immunol. Methods. 411:11-22).

Immunogen

Purified human EDN.

Application

Anti-Eosinophil-Derived Neurotoxin Antibody, clone 6D1.5/A5 is an antibody against Eosinophil-Derived Neurotoxin for use in Western Blotting, ELISA, Multiplexing.
Research Category
Inflammation & Immunology
Research Sub Category
Inflammation & Autoimmune Mechanisms
Western Blotting Analysis: 1.0 µg/mL from a representative lot detected eosinophil-derived neurotoxin in 50 µg of human liver and lung tissue lysates.
ELISA Analysis: A representative lot was employed as the capture antibody for the detection of purified human EDN, as well as EDN in serum samples from individuals with eosinophilic disorders by sandwich ELISA (Makiya, M.A., et al. (2014). J. Immunol. Methods. 411:11-22),
Multiplexing Analysis: A representative lot was employed as the capture antibody in a bead-based multiplex assay for the detection of purified human EDN as well as EDN in serum samples from healthy donors and individuals with eosinophilic disorders. Prior reduction and alkylation of the serum samples (by DTT and iodoacetamide, respectively) resulted in higher EDN levels (Makiya, M.A., et al. (2014). J. Immunol. Methods. 411:11-22),

Quality

Evaluated by Western Blotting in human spleen tissue lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected eosinophil-derived neurotoxin in 50 µg of human spleen tissue lysate.

Target description

~20 kDa observed. Target band appears larger than the calculated molecular weights of 18.35 kDa (pro-form) and 15.46 kDa (mature) due to glycosylation. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1 antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

WGK

nwg

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service