Skip to Content
MilliporeSigma
All Photos(1)

Documents

567333

Sigma-Aldrich

SNAPtide Botulinum Toxin A Substrate, Fluorogenic

≥90% (HPLC), lyophilized, FRET substrate, Calbiochem®

Synonym(s):

Botulinum Toxin Assay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.77

product name

SNAPtide Botulinum Toxin A Substrate, Fluorogenic, SNAPtide Botulinum Toxin A Substrate, Fluorogenic, is a synthetic peptide that acts as a FRET substrate with the fluorophore o-aminobenzoic acid and the acceptor chromophore 2,4-dinitrophenol (DNP).

Quality Level

Assay

≥90% (HPLC)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

fluorescence

λex ~326 nm
λem ~423 nm

shipped in

ambient

storage temp.

2-8°C

General description

A synthetic peptide, fluorescence resonance energy transfer (FRET) substrate containing the N-terminally-linked fluorophore o-aminobenzoic acid (Abz) and the acceptor chromophore 2,4-dinitrophenol (DNP) linked to the C-terminus. The substrate is based on the native botulinum toxin type A cleavage site found in the synaptosome-associated protein SNAP-25. Cleavage of the substrate by botulinum toxin releases the fluorophore and full fluorescence is restored.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Substrate for the native botulinum toxin
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from water.

Preparation Note

Prepare a 5 mM stock solution of this peptide in DMSO as follows: Add 40 µl of DMSO to a vial containing 200 nmol of peptide.

Reconstitution

Following reconstitution, aliquot, cover with foil to protect from light, and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Bigalke, H. and Shoer, L.F. 2000. In: Handbook of Experimental Pharmacology,145, Bacterial Protein Toxins (K. Aktories and I. Just, Eds.) pp. 407-443, Springer-Verlag, Berlin.
Schiavo, G., et al. 1993. J. Biol. Chem.268, 23784.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service