Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABF471

Sigma-Aldrich

Anti-Ly-6G Antibody (mouse), PerCP-Cy5.5, clone 1A8

clone 1A8, 0.2 mg/mL, from rat

Synonym(s):

Lymphocyte antigen 6G, Ly-6G, Ly-6G.1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$403.00

$403.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$403.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$403.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rat

Quality Level

conjugate

PerCP-Cy5.5

antibody form

purified antibody

antibody product type

primary antibodies

clone

1A8, monoclonal

species reactivity

mouse

concentration

0.2 mg/mL

technique(s)

flow cytometry: suitable

isotype

IgG2aκ

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Ly6G(546644)

General description

The 1A8 antibody binds to mouse Ly-6G, commonly known as Gr-1, a member of the Ly-6 superfamily of GPI-anchored cell surface proteins with roles in cell signaling and cell adhesion. Gr-1 is differentially expressed during development and maturation of cells in the myeloid lineage and is expression at varying stages and levels on monocytes, macrophages, granulocytes, and peripheral neutrophils. In the mouse, the 1A8 antibody is typically used in combination with the macrophage labeling antibody M1/70 (Anti-CD11b) for phenotypic analysis of monocytes, macrophages and granulocytes. Note: for identification of Ly-C, an alternative antibody, clone RB6-8C5, has been reported to cross-react with Ly-6C on cells expressing this antigen (Fleming et al. 1993. J. Immunol. 151:2399-2408 and Sasmono et al. 2007. J. Leukoc. Biol. 82: 111-123) and has been cited in the literature for identification of Ly-6G/Ly-6C.
This rat monoclonal antibody Anti-Ly-6G (mouse), PerCP-Cy5.5 & clone 1A8 validated for use in Flow Cytometry for the detection of Ly-6G.

Quality

The monoclonal antibody was purified from tissue culture supernatant via affinity chromatography. The purified antibody was conjugated under optimal conditions, with unreacted dye removed from the preparation.

Flow Cytometry Analysis: 0.25 µg of this antibody detected Ly-6G in one million C57Bl/6 bone marrow cells.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service