Skip to Content
MilliporeSigma
All Photos(2)

Documents

MAB1680-C

Sigma-Aldrich

Anti-Filamin A Antibody, clone TI10, Ascites Free

clone TI10, 1 mg/mL, from mouse

Synonym(s):

Filamin-A, FLN-A, Actin-binding protein 280, ABP-280, Alpha-filamin, Endothelial actin-binding protein, Filamin-1, Non-muscle filamin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

TI10, monoclonal

species reactivity

human

species reactivity (predicted by homology)

bovine (based on 100% sequence homology)

concentration

1 mg/mL

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FLNA(2316)

Related Categories

General description

Filamin is a structural protein that forms flexible cross-links between two actin filaments. Filamin is a homodimer of polypeptide chains each joined to the other at one end with an actin binding site ath the other. It is present in smooth muscle, fibroblasts, platelets and lymphocytes.

Specificity

This antibody recognizes Filamin A (actin-binding protein), both the unprocessed form (270-280 kDa), and the C-terminal (90-100 kDa) calpain cleavage fragment of Filamin A (Aakhus, 1992).

Immunogen

Human platelet protein corresponding to human Filamin A.

Application

Immunohistochemistry Analysis: A 1:50 and 1:500 dilution from a representative lot detected Filamin A in human prostate and kidney tissues, respectively.

Immunohistochemistry Analysis: A representative lot from an independent laboratory detected Filamin A in human tumor tissue sections (Bedolla, R. G., et al. (2009). Clin Cancer Research. 15(3):788-796.).

Western Blotting Analysis: A representative lot from an independent laboratory detected Filamin A in human normal and tumor tissue lysates (Bedolla, R. G., et al. (2009). Clin Cancer Research. 15(3):788-796.).

Immunoprecipitation/Western Blotting Analysis: A representative lot from an independent laboratory immunoprecipitated Filamin A from transfected HEK293T cell lysates. The immunoprecipitated sample was then subjected to Western Blotting using the same lot of antibody (Nakagawa, K., et al. (2010). Biochem. J. 427(2):237–245.).

Western Blotting Analysis: A representative lot from an independent laboratory detected Filamin A in transfected U937 cell lysate (Beekman, J. M., et al. (2008). J Immunol. 180(6):3938-3945.).

Western Blotting Analysis: A representative lot from an independent laboratory detected Filamin A in prostate cancer cell lysate. (Lin, J., et al. (2007). Int. J. Cancer. 121(12):2596–2605.).

Western Blotting Analysis: A representative lot from an independent laboratory detected Filamin A in LNCap and C4-2 whole cell lysates (Wang, Y., et al. (2007). Oncogene. 26(41):6061–6070.).

Western Blotting Analysis: A representative lot from an independent laboratory detected pulled-down human Filamin A proteins. (Lad, Y., et al. (2007). EMBO J. 26(17):3993–4004.).

Western Blotting Analysis: A representative lot from an independent laboratory detected Filamin A in lysate samples from normal bladder mucosa and muscle invasive bladder tumor (Smith, S. C., et al. (2007). Clin Cancer Res. 13(13):3803-3813.).

Western Blotting Analysis: A representative lot from an independent laboratory detected Filamin A in lysate samples from human prefontal cortical tissue (Koh, P. O., et al. (2003). Ach Gen Psychiatry. 60(3):311-319.).

Flow Cytometry Analysis: A representative lot from an independent laboratory detected Filamin A in transfected U937 cells (Beekman, J. M., et al. (2008). J Immunol. 180(6):3938-3945.).

Flow Cytometry Analysis: A representative lot from an independent laboratory detected Filamin A in LAN-1 cells (Bachmann, A. S., et al. (2006) Cancer Sci. 97(12):1359–1365.).

Immunofluorescence Analysis: A representative lot from an independent laboratory detected Filamin A in clinical patient samples (Kley, R. A., et al. (2007). Brain. 130(Pt 12):3250-3264.).

Immunocytochemistry Analysis: A representative lot from an independent laboratory detected Filamin A in LAN-1 cells (Bachmann, A. S., et al. (2006) Cancer Sci. 97(12):1359–1365.).
Research Category
Cell Structure
Research Sub Category
Cytoskeleton
This Anti-Filamin A Antibody, clone TI10, Ascites Free is validated for use in western blotting, IHC, IP, flow cytometry, immunofluorescence & ICC for the detection of Filamin A.

Quality

Evaluated by Western Blotting in human uterus tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Filamin A in 10 µg of human uterus tissue lysate.

Target description

~110 kDa observed. Phosphorylation of this protein may cause the band to shift in Western Blotting.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service