Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABF2186

Sigma-Aldrich

Anti-Zika Virus NS1 protein Antibody

purified antibody, clone 680, from mouse

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$249.00
100 μG
$625.00

$249.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μG
$249.00
100 μG
$625.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$249.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

680, monoclonal

mol wt

calculated mol wt 47 kDa

species reactivity

virus (Zika)

packaging

antibody small pack of 100 μg

technique(s)

ELISA: suitable

isotype

IgG

shipped in

dry ice

storage temp.

2-8°C

target post-translational modification

unmodified

Specificity

Clone 680 is a mouse monoclonal antibody that detects Zika virus NS1 protein.

Immunogen

His-tagged recombinant Zika Virus NS1 protein expressed in mammalian cells.

Application

Quality Control TestingEvaluated by ELISA with recombinant Zika virus NS1 antigen.ELISA Analysis: 1 μg/mL of this antibody detected recombinant Zika Virus NS1 protein.
Anti-Zika Virus NS1 protein, clone 680, Cat. No. MABF2186, is a mouse monoclonal antibody that detects Zika Virus NS1 protein and is tested in ELISA.

Target description

Genome polyprotein (UniProt: Q32ZE1) following its synthesis is cleaved into 13 different proteins in Zika virus. Zika virus is a member of the Flaviviridae family that contains a single positive sense RNA genome. Following the fusion of virus membrane with host endosomal membrane, RNA genome is released into the cytoplasm. It is initially translated as a single polyprotein, which is post-translationally cleaved into three structural proteins (C, PrM or M, and E) and seven nonstructural (NS) proteins. Six of the NS proteins (NS2A to NS5) are shown to form a replication complex on the cytoplasmic side of the endoplasmic reticulum membrane. The glycosylated NS1 that is reported to associates with lipids, forms a homodimer inside the cells and is shown to be essential for viral replication and late in infection. It is also secreted into the extracellular space as a hexameric lipoprotein particle and plays a role in immune evasion and pathogenesis via its interaction with components from both the innate and adaptive immune systems. NS1 is also a major antigenic marker for viral infection. NS2A functions in virion assembly and antagonizes the host immune response. The non-structural protein 2B is essential for the serine protease function of NS3. The serine protease NS3 has three different enzymatic activities - serine protease, NTPase and RNA helicase. The non-structural protein 4A of virus regulates the ATPase activity of the NS3 helicase while the non-structural protein 4B inhibits interferon-induced host STAT1 phosphorylation and its nuclear translocation. (Ref.: Song, H. et al. (2016). Nat. Struct. Mol. Biol. 23(5); 456-458; Bosch, I., et al. (2017). Sci. Transl. Med. 9(409); eaan1589).

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at +2°C to +8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service