Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

ABS1487

Sigma-Aldrich

Anti-P2Y12 Receptor

from rabbit, purified by affinity chromatography

Synonym(s):

P2Y purinoceptor 12, ADP-glucose receptor, ADPG-R, P2T(AC), P2Y(AC), P2Y(cyc), P2Y12 platelet ADP receptor, P2Y(ADP), SP1999

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$231.00
100 μG
$629.00

$231.00


Please contact Customer Service for Availability


Select a Size

Change View
25 μG
$231.00
100 μG
$629.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$231.00


Please contact Customer Service for Availability

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... P2RY12(64805)

General description

P2Y purinoceptor 12 (UniProt: Q9H244; also known as P2Y12, ADP-glucose receptor, ADPG-R, P2T(AC), P2Y(AC), P2Y(cyc), P2Y12 platelet ADP receptor, P2Y(ADP), SP1999) is encoded by the P2RY12 (also known as HORK3) gene (Gene ID: 64805) in human. P2Y12 is a multi-pass metabotropic receptor that is highly expressed in the platelets. Lower expression levels have been observed in the brain, lung, and adrenal glands. P2Y12 serves as a receptor for ADP and ATP coupled to G-proteins that inhibit the adenylyl cyclase second messenger system. It is not activated by UDP and UTP. It is required for normal platelet aggregation and blood coagulation. It is the predominant receptor involved in the ADP-stimulated activation of the glycoprotein IIb/IIIa receptor. Activation of the glycoprotein IIb/IIIa receptor results in enhanced platelet degranulation and thromboxane production, and prolonged platelet aggregation. P2Y12 receptor has also been reported to be expressed on microglia and is required to neuropathic pain following peripheral nerve injury. P2Y12 has seven transmembrane, four cytoplasmic, and four extracellular domains. The transmembrane helices are somewhat tilted and/or kinked. Agonist binding promotes a conformation change in the extracellular loops that leads to an inward movement of the transmembrane helices. Mutations in P2RY12 gene have been linked to bleeding disorders resulting from severe impairment of platelet response to ADP resulting in defective platelet aggregation. (Ref.: Burnstock G. (2009). Curr. Pharm. Des. 15(15);1717-35; Damman, P., et al. (2012). J. Thromb. Thrombolysis. 33(2); 143-153).

Specificity

This rabbit polyclonal antibody specifically detects P2Y12 receptor. It targets an epitope with in 13 amino acids from the extracellular domain.

Immunogen

Epitope: extracellular domain
KLH-conjugated linear peptide corresponding to 13 amino acids from the internal region of human P2Y purinoreceptor 12.

Application

Anti-P2Y12 Receptor, Cat. No. ABS1487, is a rabbit polyclonal antibody that detects P2Y purinoceptor 12 and has been tested for Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry (Paraffin) Analysis: A 1:250-1,000 dilution from a representative lot detected P2Y12 Receptor in human cerebral cortex and human tonsil tissue sections.

Western Blotting Analysis: 1 µg/mL from a representative lot detected P2Y12 Receptor in human platelet cell lysate.
Research Category
Signaling

Quality

Evaluated by Western Blotting in PC12 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected P2Y12 Receptor in PC12 cell lysate.

Target description

~50 kDa observed; 39.44 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service