Skip to Content
MilliporeSigma
All Photos(1)

Documents

MABF2050

Sigma-Aldrich

Anti-Peripheral Node Addressin (PNAd) Antibody, clone MECA-79

clone MECA-79, from rat

Synonym(s):

Peripheral Node Addressin, MECA-79, CD62L-ligand (PNAd)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

MECA-79, monoclonal

species reactivity

human, mouse

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgMκ

shipped in

ambient

target post-translational modification

unmodified

General description

Peripheral lymph node addressin (PNAd) is a mixture of glycoproteins that is expressed on high endothelial venules (HEV) and is required for lymphocyte homing. It is shown to be a ligand for CD62L (L-selectin) that is expressed on the surface of naïve and central memory T cells. PNAd is a sulfated and fucosylated glycoprotein recognized by the prototypic monoclonal antibody, MECA-79. Naïve T cells use PNAd to traffic into lymphoid organs in non-mucosal tissue sites. The interaction of PNAd with CD62L receptor is involved in tethering and rolling of lymphocytes along HEV in lymphoid tissues. Regulated appearance of PNAd+ HEVs is found during ontogeny of lymphoid tissues as well as in chronic inflammatory conditions. PNAd+ HEVs are reported to appear 24-48 h after birth in mouse peripheral lymph nodes and persist throughout life to support recirculation of lymphocytes. In humans PNAd+ HEVs are detected in fetal peripheral lymph nodes. PNAd+ HEVs are also shown to be present in several chronic inflammatory conditions, including thyroiditis, ulcerative colitis, psoriasis, rheumatoid synovitis, and cutaneous lymphomas. In murine models of inflammation, PNAd+ HEV-like vessels develop and these vessels are found to support lymphocyte recruitment. Studies on cancer in murine models and in humans have identified PNAd as a biomarker of improved disease prognosis and blockade of PNAd or its ligand, L-selectin, is shown to abrogate protective antitumor immunity in murine models. (Ref.: Sinha, RK et al. (2006). Vet. Immunol. Immunopathol. 110 (1-2): 97-108).

Specificity

Clone MECA-79 detects Peripheral Node Addressin (PNAd) in human and mouse.

Immunogen

Collagenase-dispersed lymph node stromal cells from BALB/c mice.

Application

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Peripheral Node Addressin (PNAd) in human liver and human pancreas tissues.

Western Blotting Analysis: A representative lot detected Peripheral Node Addressin (PNAd) in Western Blotting applications (Hemmerich, S., et. al. (1994). J Exp Med. 180(6):2219-26).

Inhibition Analysis: A representative lot detected Peripheral Node Addressin (PNAd) in Inhibition applications (Streeter, P.R., et. al. (1988). J Cell Biol. 107(5):1853-62;Michie, S.A., et. al. (1993). Am. J. Pathol. 143(6):1688-98; Berg, E.L., et. al. (1991). J Cell Biol. 114(2):343-9).

Immunoprecipitation Analysis: A representative lot detected Peripheral Node Addressin (PNAd) in Immunoprecipitation applications (Berg, E.L., et. al. (1991). J Cell Biol. 114(2):343-9).

Immunohistochemistry Analysis: A representative lot detected Peripheral Node Addressin (PNAd) in Immunohistochemistry applications (Sinha, R.K., et. al. (2006). Immunology. 119(4):461-9; Streeter, P.R., et. al. (1988). J Cell Biol. 107(5):1853-62; Michie, S.A., et. al. (1993). Am J Pathol. 143(6):1688-98).

Flow Cytometry Analysis: A representative lot detected Peripheral Node Addressin (PNAd) in Flow Cytometry applications (Sinha, R.K., et. al. (2006). Immunology. 119(4):461-9).
Research Category
Inflammation & Immunology
This rat monoclonal Anti-Peripheral Node Addressin (PNAd) Antibody, clone MECA-79, Cat. No. MABF2050, is tested in Flow Cytometry, Immunohistochemistry (Paraffin), , Immunoprecipitation, Inhibition, and Western Blotting for the detection of Peripheral node addressin.

Quality

Evaluated by Immunohistochemistry in human tonsil tissue.

Immunohistochemistry Analysis: A 1:50 dilution of this antibody detected Peripheral Node Addressin (PNAd) in human tonsil tissue.

Target description

50-250 kDa calculated.

Physical form

Format: Purified
Purified rat monoclonal antibody IgM in PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ricardo A Chaurio et al.
Immunity, 55(1), 115-128 (2022-01-13)
The immune checkpoint receptor PD-1 on T follicular helper (Tfh) cells promotes Tfh:B cell interactions and appropriate positioning within tissues. Here, we examined the impact of regulation of PD-1 expression by the genomic organizer SATB1 on Tfh cell differentiation. Vaccination

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service