Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABF1085

Sigma-Aldrich

Rat IgG2b Negative Control, clone LTF-2 Antibody

clone LTF-2, from rat, purified by affinity chromatography

Synonym(s):

Immunoglobulin G2b, IgG2b

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$128.00

$128.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$128.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.42

$128.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rat

Quality Level

antibody form

affinity isolated antibody

clone

LTF-2, monoclonal

purified by

affinity chromatography

technique(s)

flow cytometry: suitable

isotype

IgG2b

target post-translational modification

unmodified

General description

The LTF-2 immunoglobulin is useful as an isotype-matched control. The LTF-2 immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG2b antibodies.

Immunogen

Immunogen information for this antibody is proprietary information.

Application

Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
This rat monoclonal Rat IgG2b Negative Control, clone LTF-2, Cat. No. MABF1085 is validated for use in Flow Cytometry.

Physical form

Affinity Chromatography
Purified Rat monoclonal containing 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH7.2. This monoclonal antibody preparation was purified from tissue culture supernatant via affinity chromatography.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.
Note: It is recommended to store the product undiluted at 2-8°C and protected from prolonged exposure to light. Do not freeze.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service