Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABE241

Sigma-Aldrich

Anti-dimethyl-phospho Histone H3 (Lys9/27)/(Ser10/28) Antibody, clone 51TA-2H12

ascites fluid, clone 51TA-2H1, from mouse

Synonym(s):

Histone H3.1, Histone H3/a, Histone H3/b, Histone H3/c, Histone H3/d, Histone H3/f, Histone H3/h, Histone H3/I, Histone H3/j, Histone H3/k, Histone H3/l

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$669.00

$669.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$669.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$669.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

51TA-2H1, monoclonal

species reactivity

human

technique(s)

dot blot: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer10/pSer28), dimethylation (Lys9/Lys27)

Gene Information

human ... HIST1H3F(8968)

General description

Histones are nuclear proteins that form octameric structures which bind DNA to form units of chromatin called nucleosomes. The family of histones—H2A, H2B, H3, and H4—are key players in gene regulation. They undergo a number of post-translational modifications (PTM) in response to various stimuli, including phosphorylation on serine and threonine residues and methylation on lysine residues. PTMs produce configural changes in histone proteins that may induce nucleosome remodeling and expose or hide DNA sequences from transcriptional complexes. Methylation occurs on histone H3 at lysine 4, 9, 27, 36, and 79 residues. Previous studies have reported that methylation of H3 lysine 9 provides a binding site for heterochromatin adaptor proteins (HP1) which are involved in the formation of heterochromatin and gene repression. Conversely,deacetylation of histone H3 lysine 9 is associated with gene-activating events. Often, it is the pattern of different modifications on several histone residues that alter chromatin, rather than modification of single residues.

Specificity

This antibody recognizes Histone H3 dimethylated and phosphorylated at Lys9/27 and Ser10/28.

Immunogen

Epitope: Dimethylated and phosphorylated at Lys9/27 and Ser10/28.
Ovalbumin-conjugated linear peptide corresponding to human Histone H3 dimethylated and phosphorylated at Lys9/27 and Ser10/28.

Application

Anti-dimethyl-phospho Histone H3 (Lys9/27)/(Ser10/28) Antibody, clone 51TA-2H12 is a Mouse Monoclonal Antibody for detection of dimethyl-phospho Histone H3 (Lys9/27)/(Ser10/28) also known as Histone H3.1 & has been validated in WB, DB, ICC, IP & IHC.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
Western Blot Analysis: A representative lot was used by an independent laboratory in KCl whole-cell extract. (Eberlin, A., et al. (2008). Molecular and Cellular Biology. 28(5):1739–1754.)

Dot Blot Analysis: A representative lot was used by an independent laboratory in unmodified and modified Histone H3 peptides. (Eberlin, A., et al. (2008). Molecular and Cellular Biology. 28(5):1739–1754.)

Immunocytochemistry Analysis: A representative lot was used by an independent laboratory in NIH/3T3 cells. (Eberlin, A., et al. (2008). Molecular and Cellular Biology. 28(5):1739–1754.)

Immunohistochemistry Analysis: A representative lot was used by an independent laboratory in mouse testis tissue. (Eberlin, A., et al. (2008). Molecular and Cellular Biology. 28(5):1739–1754.)

Immunoprecipitation Analysis: A representative lot was used by an independent laboratory in HeLa cells. (Eberlin, A., et al. (2008). Molecular and Cellular Biology. 28(5):1739–1754.)

Quality

Evaluated by Western Blot in untreated and etoposide acid extract treated HeLa cell lysates.

Western Blot Analysis: A 1:5,000 dilution of this antibody detected Histone H3 on 10 µg of untreated and etoposide acid extract treated HeLa cell lysates.

Target description

~17 kDa observed

Physical form

Mouse monoclonal IgG1κ ascites with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Untreated and etoposide acid extract treated HeLa cell lysates.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service