Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABE263-AF647

Sigma-Aldrich

Anti-unmethylated Histone H3 (Lys9) Antibody, clone 9B1-2G6, Alexa Fluor 647 Conjugate

clone 9B1-2G6, from mouse, ALEXA FLUOR 647

Synonym(s):

Histone H3.1, Histone H3/a, Histone H3/b, Histone H3/c, Histone H3/d, Histone H3/f, Histone H3/h, Histone H3/I, Histone, H3/j, Histone H3/k, Histone H3/l

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 647

antibody form

purified antibody

antibody product type

primary antibodies

clone

9B1-2G6, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Gene Information

human ... HIST1H3F(8968)

General description

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the ′beads on a string′ structure. Histone proteins are highly post-translationally modified however Histone H3 is the most extensively modified of the five histones. Histone H3 sequence variants and variable modification states are thought to play a role in the dynamic and long term regulation of genes. Trimethylation of histone H3 on Lys9 (H3K9me3) is one of the most highly studied epigenetic marks. H3K9me3 functions in the repression of euchromatic genes, and in epigenetic control of heterochromatin assembly, most likely by acting as a recognition motif for the binding of chromatin-associated proteins, such as Swi6 or HP1α. The enzymes responsible for H3K9 trimethylation are SUV39H1 and SUV39H2. While lysine demethylation is mediated by enzymes such as JmjC domain-containing proteins.

Specificity

This antibody recognizes Histone H3 unmethylated at Lys9. This antibody does not cross react with methyl-Histone (Lys29). This antibody detects unmethylated Histone H3 (Lys9) when Ser10 is phosphorylated.

Immunogen

KLH-conjugated linear peptide corresponding to human unmethylated Histone H3 (Lys9).

Application

Research Category
Epigenetics & Nuclear Function

Epigenetics & Nuclear Function
Research Sub Category
Histones

Chromatin Biology
See data sheet for Cat. No. MABE263, Anti-unmethylated Histone H3 (Lys9), clone 9B1-2G6, for additional Western blot and dot blot specificity analysis performed using an unconjugated version of this antibody.
This Anti-unmethylated Histone H3 (Lys9) Antibody, clone 9B1-2G6 is validated for use in Immunocytochemistry for the detection of unmethylated Histone H3 (Lys9).

Quality

Immunocytochemistry Analysis:

A 1:50 dilution of this antibody detected unmethylated Histone H3 (Lys9) in HeLa cells.
Alexa Fluor is a registered trademark of Life Technologies.

Target description

~15 kDa observed. Molecular weight was observed in parent product MABE263

Physical form

Protein G Purified
Purified conjugated mouse monoclonal antibody in buffer containing PBS and 15 mg/ml BSA with 0.1% Sodium Azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service