Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABF1980

Sigma-Aldrich

Anti-Complement receptor C5aR (CD88) Antibody, clone S5/1

clone S5/1, from mouse

Synonym(s):

C5a anaphylatoxin chemotactic receptor 1, C5a anaphylatoxin chemotactic receptor, C5a-R, CD88

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$231.00
100 μL
$607.00

$231.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
$231.00
100 μL
$607.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

$231.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

S5/1, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μL

technique(s)

ELISA: suitable
activity assay: suitable
flow cytometry: suitable
immunoprecipitation (IP): suitable
inhibition assay: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... C5AR1(728)

General description

C5a anaphylatoxin chemotactic receptor 1 (UniProt: P21730; also known as C5a anaphylatoxin chemotactic receptor, C5a-R, C5aR, CD88) is encoded by the C5AR1 (also known as C5AR, C5R1) gene (Gene ID: 728) in human. C5aR is a multi-pass membrane protein that serves as a receptor for the chemotactic and inflammatory peptide anaphylatoxin C5a. It exists as a homodimer, but can also form higher-order oligomers. C5a interacts with at least two sites on the receptor: a high-affinity site on the extracellular N-terminus, and a second site in the transmembrane region, which activates downstream signaling events. Receptor activation stimulates chemotaxis, granule enzyme release, and intracellular calcium release and superoxide anion production. Chemotaxis inhibitory protein of Staphylococcus aureus (CHIPS), a 121-residue protein, is reported to bind to the N-terminus of the C5aR (aa 1-35) with nanomolar affinity and inhibits C5a-mediated responses in human leukocytes. Sulfation at Tyrosine 14 is shown to be important for CHIPS binding. Upon C5a binding, C5aR) undergoes rapid phosphorylation on the six serine residues present in the C-terminal region followed by desensitization and internalization. The key residues involved in this process are Serine 334 and Serine 338. Phosphorylated C5aR is shown to co-localize with beta-arrestin 1 and 2 in cytoplasmic vesicles. Although C5aR activation is able to promote a loose association with beta-arrestins, but phosphorylation of either Serine 334)/338 or Serine 327)/338 is reported to be essential for the formation of a persistent complex. (Ref.: Ippel, JH., et al., (2009). J. Biol. Chem. 284(18); 12363 12372; Braun, L., et al. (2003). J. Biol. Chem. 278(6); 4277-4285).

Specificity

Clone S5/1 specifically detects C5a anaphylatoxin chemotactic receptor 1 (C5aR). It targets an epitope within the first 31 amino acids from the N-terminal region.

Immunogen

BSA-conjugated linear peptide corresponding to the first 31 amino acids from the N-terminal region of human C5a anaphylatoxin chemotactic receptor 1.
Epitope: N-terminus

Application

Anti-Complementreceptor C5aR (CD88), clone S5/1, Cat. No. MABF1980 is a mouse monoclonal antibody that detects C5a anaphylatoxin chemotactic receptor 1 and has been tested for use in ELISA, Flow Cytometry, Immunoprecipitation, Inhibition/Activity assays, andn Western Blotting.
ELISA Analysis: A representative lot detected Complementreceptor C5aR (CD88) in ELISA applications (Pollok-Kopp, B., et. al. (2007). J Biol Chem. 282(7):4345-53).

Inhibits Activity/Function Analysis: A representative lot detected Complementreceptor C5aR (CD88) in Inhibits Activity applications (Oppermann, M., et. al. (1993). J Immunol. 151(7):3785-94).

Flow Cytometry Analysis: A representative lot detected Complementreceptor C5aR (CD88) in Flow Cytometry applications (Oppermann, M., et. al. (1993). J Immunol. 151(7):3785-94; Pollok-Kopp, B., et. al. (2007). J Biol Chem. 282(7):4345-53).

Immunoprecipitation Analysis: A representative lot detected Complementreceptor C5aR (CD88) in Immunoprecipitation applications (Pollok-Kopp, B., et. al. (2007). J Biol Chem. 282(7):4345-53).
Research Category
Inflammation & Immunology

Quality

Evaluated by Western Blotting in two BSA-conjugated C5aR peptides (aa 1-31 and 327-344) phosphorylated at Serine 332, 334, and 338.

Western Blotting Analysis: A 1:20,000 dilution of this antibody detected Complementreceptor C5aR (CD88) in two BSA-conjugated C5aR peptides (aa 1-31 and 327-344) phosphorylated at Serine 332, 334, and 338

Target description

~100-110 kDa observed; 39.34 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2a in PBS without azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C5aR1 signaling triggers lung immunopathology in COVID-19 through neutrophil extracellular traps.
Silva, et al.
The Journal of Clinical Investigation (2023)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service