Skip to Content
MilliporeSigma
All Photos(4)

Documents

SAB5900203

Sigma-Aldrich

Alpaca Anti-hTNFR, recombinant single domain Antibody

new

Recombinant Alpaca(VHH) Nano antibody

Synonym(s):

CD120a, TNFR1, tumor necrosis factor receptor superfamily member 1A, tumor necrosis factor receptor type 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

alpaca

Quality Level

recombinant

expressed in E. coli

conjugate

unconjugated

antibody form

affinity purified VHH antibody

clone

TR09, monoclonal

Assay

>95% (SDS-PAGE)

form

lyophilized solid

mol wt

14.2 kDa
~15-18 kDa (SDS PAGE under reducing condition)

purified by

immobilized metal affinity chromatography (IMAC)

species reactivity

human

concentration

1 mg/mL

technique(s)

ELISA: 1:1000-1:10000
immunofluorescence: 1:100-1:300
immunohistochemistry: 1:100-1:300
western blot: 1:500-1:3000

color

colorless

pH

( in PBS, pH 7.4)

isotype

VHH

Protein ID accession no.

UniProt accession no.

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... TNFRSF1A(7132)

General description

Tumor necrosis factor receptor 1 (TNFR1), also known as tumor necrosis factor receptor superfamily member 1A (TNFRSF1A) and CD120a (cluste of differentiation 120a), is an ubiquitous membrane receptor that binds tumor necrosis factor-alpha (TNFα). It has been shown to be localized to both plasma membrane lipid rafts and the trans golgi complex with the help of the death domain (DD). TNFR1 can activate the transcription factor NF-κB, mediate apoptosis, and regulate inflammation processes.Alpaca derived anti-TNFR1 VHH single domain antibody (molecular weight: 14.2 kDa), expressed in E. coli under conditions free from animal derived components.It has a 6His tag at its C-terminal.
Single-domain antibody (sdAb), also called nano-antibody, is the variable region isolated from the antigen-binding variable domain (VHH) of camelid heavy-chain-only antibody. It lacks light chains and a first constant domain(CH1). This configuration confers a number of unique advantages over conventional monoclonal antibodies. The small size of just 15 kDa (2~4 nm), single-domain antibody is the smallest antigen-binding fragment that can easily penetrate tissues, allowing them to bind epitopes that are inacessible to conventional antibodies. These single-domain antibodies offer superior chemical and thermal stability and increased solubility than conventional antibodies. All our antibodies are tested to ensure the highest level of specificity, sensitivity and reproducibility.

Specificity

Human Tumor necrosis factor receptor 1 (TNFR1) / CD120a

Immunogen

Recombinant human TNFR1 (Ile22-Thr211) protein

Application

ELISA, WB, IHC, IF

Packaging

50 µg per vial / 100 µg per vial

Physical form

Affinity purified VHH single domain antibody lyophilized from PBS, pH7.6

Reconstitution

Reconstitute the lyophilized antibody with sterile deionized water or with 50% glycerol (ACS grade or better) to a stock solution of 1 mg/mL. Solubilize for 30 to 60 minutes at room temperature with occasional gentle mixing. Avoid vigorous shaking or vortexing.

Storage and Stability

Stable for 1 year at 4°C from date of receipt.

Other Notes

Storage at 4°C; Do not freeze.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service