Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABC182

Sigma-Aldrich

Anti-NRF-1 Antibody, clone 10B11.1

clone 10B11.1, from mouse

Synonym(s):

Nuclear respiratory factor 1, Alpha-pal, Alpha palindromic-binding protein, NRF-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

10B11.1, monoclonal

species reactivity

mouse

species reactivity (predicted by homology)

human (based on 100% sequence homology)

technique(s)

western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... NRF1(4899)

General description

Nuclear respiratory factor 1 (UniProt Q16656; also known as Alpha-pal, Alpha palindromic-binding protein, NRF-1) is encoded by the NRF1 (Also known as ALPHA-PAL) gene (Gene ID 4899) in human. NRF-1 is a redox-responsive transcription factor that plays an anti-oxidant, pro-survival role in mitochondrial biogenesis process. NRF-1 is upregulated and activated/phosphorylated by Akt in response to oxidant exposure or increased cellular reactive oxygen species (ROS), resulting in NRF-1 nuclear translocation, where it binds to DNA as a homodimer and mediates the transcription of mitochondrial transcription factor A (mtTFA) that is needed for mitochondrial DNA transcription.

Specificity

Immunogen construct region is present in human NRF-1 spliced isoform Long, isfoform Short, and isfoform 4, but is more than 40% absent from spliced isoform 4 reported by UniProt (Q16656).

Immunogen

Epitope: Internal (N-terminal half).
GST-tagged recombinant human NRF-1 internal fragment.

Application

Research Category
Apoptosis & Cancer
Research Sub Category
Transcription Factors
This Anti-NRF-1 Antibody, clone 10B11.1 is validated for use in Western Blotting for the detection of NRF-1.

Quality

Evaluated by Western Blotting in NIH/3T3 cell lysate.

Western Blotting Analysis: 0.25 µg/mL of this antibody detected NRF-1 in 10 µg of NIH/3T3 cell lysate.

Target description

~58 kDa observed. 53.54/53.57 kDa (human/mouse isoform Long), 46.16 kDa (human/mouse isoform Short), 36.05 kDa (human isoform 3), 55.67 kDa (human isoform 4) calculated.

Physical form

Format: Purified
Protein G purified.
Purified mouse monoclonal IgG2bκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service