Skip to Content
MilliporeSigma
All Photos(1)

Documents

ST1204

Sigma-Aldrich

Anti-PGC-1α Rabbit pAb

liquid, Calbiochem®

Synonym(s):

PGC-1α Detection Antibody, Rabbit Anti-PGC-1α

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

does not contain

preservative

species reactivity

human, mouse, rat

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−70°C

target post-translational modification

unmodified

Gene Information

General description

Protein G purified rabbit polyclonal antibody. Recognizes the ~113 kDa PGC-1α protein.
Recognizes the ~113 kDa PGC-1α protein in BAT cells.
This Anti-PGC-1α Rabbit pAb is validated for use in Immunoblotting, Immunoprecipitation for the detection of PGC-1α.

Immunogen

A recombinant protein consisting of amino acids 1-120 of mouse PGC-1α

Application


Immunoblotting (not recommended)
Immunoprecipitation (1-2 g/100-200 g total protein)

Warning

Toxicity: Regulatory Review (Z)

Physical form

In 50 mM PBS

Analysis Note

Positive Control
Brown adipose tissue from mice exposed to cold

Other Notes

Chang, J.S., et al. 2010. J. Biol. Chem. 285, 18039.
Zhang, Y., et al. 2009. J. Biol. Chem. 284, 32813.
Lai, L., et al. 2008. Genes Dev. 14, 1948.
Rodgers, J.T., et al. 2008. FEBS Lett. 582, 46.
Mazzucotelli, A., et al. 2007. Diabetes 10, 2467.
Nemoto, S., et al. 2005. J. Biol. Chem. 16, 16456.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Santosh Bhat et al.
Circulation. Heart failure, 12(3), e005529-e005529 (2019-02-26)
Proper dynamics of RNA polymerase II, such as promoter recruitment and elongation, are essential for transcription. PGC-1α (peroxisome proliferator-activated receptor [PPAR]-γ coactivator-1α), also termed PPARGC1a, is a transcriptional coactivator that stimulates energy metabolism, and PGC-1α target genes are downregulated in
Kennedy C Whitley et al.
Physiological reports, 8(14), e14517-e14517 (2020-07-31)
Calcineurin is a Ca2+ -dependent serine/threonine phosphatase that dephosphorylates nuclear factor of activated T cells (NFAT), allowing for NFAT entry into the nucleus. In skeletal muscle, calcineurin signaling and NFAT activation increases the expression of proliferator-activated receptor-gamma coactivator 1-alpha (PGC-1α)
Elizabeth A Novak et al.
Frontiers in immunology, 14, 1231700-1231700 (2023-09-25)
We have previously demonstrated that a pathologic downregulation of peroxisome proliferator-activated receptor-gamma coactivator 1-alpha (PGC1α) within the intestinal epithelium contributes to the pathogenesis of inflammatory bowel disease (IBD). However, the mechanism underlying downregulation of PGC1α expression and activity during IBD
Ryan W Baranowski et al.
iScience, 26(7), 107047-107047 (2023-06-26)
We examined the effects of ∼30 days of spaceflight on glycogen synthase kinase 3 (GSK3) content and inhibitory serine phosphorylation in murine muscle and bone samples from four separate missions (BION-M1, rodent research [RR]1, RR9, and RR18). Spaceflight reduced GSK3β content
Valerio Donato et al.
Nature cell biology, 19(4), 341-351 (2017-03-21)
Self-renewing naive mouse embryonic stem cells (mESCs) contain few mitochondria, which increase in number and volume at the onset of differentiation. KBP (encoded by Kif1bp) is an interactor of the mitochondrial-associated kinesin Kif1Bα. We found that TDH, responsible for mitochondrial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service