Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB8701

Sigma-Aldrich

Anti-Dengue Virus Type I Antibody, clone 15F3-1

clone 15F3-1, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$309.00

$309.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$309.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$309.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

clone

15F3-1, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

isotype

IgG1

shipped in

wet ice

General description

Dengue fever is an acute, mosquito-transmitted viral disease characterized by fever, headache, arthralgia (severe retro-orbital pain), myalgia, rash, nausea, and vomiting. Infections are caused by any of the four closely related, but antigenically distinct virus serotypes (DEN-1, DEN-2, DEN-3, and DEN-4). Infection with one of these serotypes does not provide cross-protective immunity, so persons living in a dengue-endemic area can have four dengue infections during their lifetimes. Dengue is primarily an urban disease of the tropics, and the viruses that cause it are maintained in a cycle that involves humans and Aedes aegypti, a domestic, day-biting mosquito that prefers to feed on humans. Although most dengue infections result in relatively mild illness, some can produce Dengue Hemorrhagic Fever (DHF) or dengue shock syndrome, with children being particularly at risk. Although epidemic outbreaks have been reported since 1779, the incidence has been increasing, with global, multiple serotype pandemics intensifying within the last 15 years. There is no specific antiviral therapy for dengue, but for both classical dengue and dengue hemorrhagic fever, symptomatic and supportive measures are effective. Important risk factors for DHF include the strain and serotype of the virus involved, as well as the age, immune status, and genetic predisposition of the patient.

Specificity

Reacts with the Dengue type 1 virus. May show reactivity with Dengue type 2 virus.

Immunogen

Dengue type 1 (Hawaiian) virus antigen.

Application

Anti-Dengue Virus Type I Antibody, clone 15F3-1 detects level of Dengue Virus Type I & has been published & validated for use in IF.
Immunofluorescent assay. Final working dilutions must be determined by end user.

Physical form

0.02M Phosphate Buffer, pH 7.6, 0.25M NaCl and 0.1% sodium azide.
Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sambashiva Banala et al.
ACS chemical biology, 8(10), 2127-2132 (2013-08-15)
Detection of antibodies is essential for the diagnosis of many diseases including infections, allergies, and autoimmune diseases. Current heterogeneous immunoassays require multiple time-consuming binding and washing steps, which limits their application in point-of-care diagnostics and high-throughput screening. Here, we report

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service