Skip to Content
MilliporeSigma
All Photos(14)

Documents

ZRB1939

Sigma-Aldrich

Anti-IGF-2R/CD222 Antibody, clone 2O9 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Cation-independent mannose-6-phosphate receptor;CI Man-6-P receptor;CI-MPR;M6PR;300 kDa mannose 6-phosphate receptor;MPR 300;Insulin-like growth factor 2 receptor;Insulin-like growth factor II receptor;IGF-II receptor;M6P/IGF2 receptor;M6P/IGF2R;CD222

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2O9, recombinant monoclonal

description

2O9 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 274.38 kDa
observed mol wt ~275 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... IGF2R(3482)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2O9 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects IGF-2R/CD222. It targets an epitope within 20 amino acids from the C-terminal half, lumenal domain.

Immunogen

KLH-conjugated linear peptide corresponding to 20 amino acids from the C-terminal half of human IGF-2R/CD222.

Application

Quality Control Testing

Evaluated by Western Blotting in SK-MEL-28 cell lysates.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected IGF-2R/CD222 in SK-MEL-28 cell lysates.

Tested Applications

Western Blotting Analysis: A 1:10,000 dilution from a representative lot detected IGF-2R/CD222 in Jurkat and HepG2 cell lysates.

Affinity Binding Assay: A representative lot of this antibody bound IGF-2R/CD222 peptide with a KD of 3.7 x 10-6 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected IGF-2R/CD222 in SK-MEL-28 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected IGF-2R/CD222 in human placenta tissue sections.

Flow Cytometry Analysis: 0.1 μg from a representative lot detected IGF-2R/CD222 in one million HepG2 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Cation-independent mannose-6-phosphate receptor (UniProt: P11717; also known as CI Man-6-P receptor, CI-MPR, M6PR, 300 kDa mannose 6-phosphate receptor, MPR 300, Insulin-like growth factor 2 receptor, Insulin-like growth factor II receptor, IGF-II receptor, M6P/IGF2 receptor, M6P/IGF2R, CD222) is encoded by the IGF2R (also known as MPRI) gene (Gene ID: 3482) in human. IGF-2R/CD222 is a single-pass type I membrane protein found in Golgi and endosome. It is synthesized with a signal peptide (aa 1-40), which is subsequently cleaved off to produce the mature protein that contains a long lumenal domain (aa 41-2304), a transmembrane domain (aa 2305-2327), and a cytoplasmic domain (aa 2328-2491). It contains 15 mannose 6-phosphate receptor homology (MRH) domains in its extracellular region. IGF-2R/CD222 mediates the transport of phosphorylated lysosomal enzymes from the Golgi complex and the cell surface to lysosomes. Lysosomal enzymes bearing phosphomannosyl residues bind specifically to mannose-6-phosphate receptors (MPR) in the Golgi apparatus and the resulting receptor-ligand complex is transported to an acidic pre-lysosomal compartment where the low pH mediates the dissociation of the complex. The receptor is then recycled back to the Golgi for another round of trafficking through its binding to the retromer. IGF-2R/CD222 undergoes cysteine-S-palmitoylation that promotes interaction with the retromer cargo-selective complex which mediates its retrograde trafficking to the Golgi apparatus. IGF-2R/CD222 is reported to transport Lck intracellularly and direct its distribution to CD45 at the plasma membrane. The loss of IGF-2R/CD222 results in a decrease in signal transduction and subsequently a blockade of T cell effector functions. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Pfisterer, K., et al. (2014). J. Immunol.193(6); 2718-2732; McCormick, PJ., et al. (2008). Traffic. 9(11); 1984-1997).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service