Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB5240

Sigma-Aldrich

Anti-Phosphacan Antibody

culture supernatant, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

monoclonal

species reactivity

chicken

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgM

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... PTPRZ1(5803)

General description

Phosphacan is a soluble nervous tissue-specific proteoglycan that is thought to regulate axonal outgrowth. It is synthesized by glia and binds to neurons and to the neural cell adhesion molecules tenascin, N-CAM or NG-CAM but not to laminin and fibronectin. Phosphacan acts as a potent inhibitor of cell adhesion and neurite outgrowth.

Specificity

Phosphacan (Receptor Tyrosine Phosphatase Beta).

Immunogen

Mixture of chicken chondroitin sulfate proteoglycans.

Application

Detect Phosphacan using this Anti-Phosphacan Antibody validated for use in ELISA, WB, IH(P).
Research Category
Neuroscience
Research Sub Category
Growth Cones & Axon Guidance
Western blot: 1:1,000

Samples must be digested with chondroitinase prior to running on SDS gels because undigested phosphacan is too large for most gels. Treatment is at a concentration of chondroitinase of 10U/mL in Tris-HCL pH 8.0. Make tissue or cell extract in 20-50mM Tris pH 7.6-8.0 with 0.15M NaCl in the presence of protease inhibitors. Add 1 microliter of enzyme to 30 microliters of extract and incubate 30 minutes at 37C. Then add SDS sample buffer, heat or boil sample as normal for SDS reducing samples.

Immunohistochemistry: 1:20 Works on cold methanol or paraformaldehyde fixed tissues. Works on paraffin embedded tissues. Chondrotinase treatment of tissue consists of treating mounted tissue section with a solution of PBS plus 0.5U/mL of Chondroitinase ABC for 30 minutes at room temperature prior to blocking and antibody staining.

Immunocytochemistry: not tested

ELISA: Excellent for core and monomer protein.

Optimal working dilutions must be determined by the end user.

Physical form

Tissue culture supernatant. Liquid. Contains no preservative.

Storage and Stability

Maintain at -20°C in undiluted aliquots up to 6 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service