Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABN282

Sigma-Aldrich

Anti-CNGB1 Antibody, clone 5A5.1

clone 5A5.1, 1 mg/mL, from mouse

Synonym(s):

Cyclic nucleotide-gated cation channel beta-1, Cyclic nucleotide-gated cation channel 4, CNG channel 4, CNG-4, CNG4, Cyclic nucleotide-gated cation channel gamma, Cyclic nucleotide-gated cation channel modulatory subunit, Cyclic nucleotide-gated channel

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$532.00

$532.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$532.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$532.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5A5.1, monoclonal

species reactivity

human

concentration

1 mg/mL

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CNGB1(1258)

General description

CNGB1, also known as Cyclic nucleotide-gated cation channel beta-1, Cyclic nucleotide-gated cation channel 4 (CNG4), Cyclic nucleotide-gated cation channel gamma, Cyclic nucleotide-gated cation channel modulatory subunit, Cyclic nucleotide-gated channel beta-1, or Glutamic acid-rich protein (GARP), and encoded by the gene CNGB1/CNCG2/CNCG3L/CNCG4/RCNC2, is a subunit of cyclic nucleotide-gated (CNG) channels which are nonselective cation channels expressed in retinal and olfactory neurons. The CNG channels play important roles in both visual and olfactory signal transduction. When associated with CNGA1, CNGB1 is involved in the regulation of ion flow into the rod photoreceptor outer segment (ROS), in response to light-induced alteration of the levels of intracellular cGMP. CNGB1 is localized in the plasma membranes of expressing cells and mediates the ion transport of the channel. Mutations in CNGB1 are associated with the genetic disease retinitis pigmentosa 45 that is a progressive vision disease associated with the loss of rod and cone photoreceptors.

Immunogen

GST-tagged recombinant protein corresponding to human CNGB1.

Application

Detect CNGB1 using this Anti-CNGB1 Antibody, clone 5A5.1 validated for use in western blotting.
Research Category
Neuroscience
Research Sub Category
Developmental Signaling

Quality

Evaluated by Western Blotting in Y79 cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected CNGB1 in 10 µg of Y79 cell lysate.

Target description

~140 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anastasia C Hepburn et al.
Cancers, 13(15) (2021-08-08)
Cisplatin-based neoadjuvant chemotherapy (NAC) is recommended prior to radical cystectomy for muscle-invasive bladder cancer (MIBC) patients. Despite a 5-10% survival benefit, some patients do not respond and experience substantial toxicity and delay in surgery. To date, there are no clinically

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service