Skip to Content
MilliporeSigma
All Photos(1)

Documents

MAB3776

Sigma-Aldrich

Anti-MDM2 Antibody, clone SMP14

clone SMP14, Chemicon®, from mouse

Synonym(s):

Murine Double Minute 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

SMP14, monoclonal

species reactivity

rat, human, mouse

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation (IP): suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MDM2(4193)
mouse ... Mdm2(17246)
rat ... Mdm2(314856)

General description

MDM2 is over-expressed in many tumors. Its principal function is the ubiquitination and degradation of p53 tumour suppressor protein. This monoclonal antibody also recognizes a peptide epitope around Thr-216 of murine MDM2 when Thr-216 is unphosphorylated (Zhang & Prives, 2001). SMP14 also cross reacts with some cytokeratins (6, 14 & 16). This is only a problem when working with certain epithelial cells and not fibroblasts.

Specificity

Recognizes human MDM2.

Immunogen

Synthetic peptide corresponding to residues 154-167 (CSRPSTSSRRRAISE) of human MDM2 cross-linked to KLH using glutaldehyde.

Application

Research Sub Category
Ubiquitin & Ubiquitin Metabolism

RNA Binding Protein (RBP)
Use Anti-MDM2 Antibody, clone SMP14 (Mouse Monoclonal Antibody) validated in IP, IHC, IHC(P) to detect MDM2 also known as Murine Double Minute 2.
Western blot

Immunohistochemistry (Frozen and Paraffin-embedded sections)

Immunoprecipitation

ELISA

Optimal working dilutions must be determined by end user.

Physical form

Format: Purified
Liquid in 0.02M phosphate buffer, 0.25M NaCl, pH 7.6, with 0.1% sodium azide

Analysis Note

Control
POSITIVE CONTROL: Breast carcinoma

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gengze Wu et al.
Circulation, 130(17), 1452-1465 (2014-08-27)
Long noncoding RNAs (lncRNAs) have recently been implicated in many biological processes and diseases. Atherosclerosis is a major risk factor for cardiovascular disease. However, the functional role of lncRNAs in atherosclerosis is largely unknown. We identified lincRNA-p21 as a key
Richard O Adeyemi et al.
Journal of virology, 86(15), 8328-8332 (2012-05-25)
The DNA damage response to infection with minute virus of mice (MVM) leads to activated p53; however, p21 levels are reduced via a proteasome-mediated mechanism. This loss was sustained, as virus replicated in infected cells held at the G(2)/M border.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service