MilliporeSigma
  • Replication of minute virus of mice in murine cells is facilitated by virally induced depletion of p21.
Replication of minute virus of mice in murine cells is facilitated by virally induced depletion of p21.
Journal of virology (2012-05-25)
Richard O Adeyemi, David J Pintel
ABSTRACT
The DNA damage response to infection with minute virus of mice (MVM) leads to activated p53; however, p21 levels are reduced via a proteasome-mediated mechanism. This loss was sustained, as virus replicated in infected cells held at the G(2)/M border. Addition of the cyclin-dependent kinase (CDK) inhibitor roscovitine after S-phase entry reduced MVM replication, suggesting that CDK activity was critical for continued viral replication and virus-induced reduction of p21 may thus be necessary to prevent inhibition of CDK.
Sigma-Aldrich
Roscovitine, ≥98% (TLC)
Sigma-Aldrich
Anti-MDM2 Antibody, clone SMP14, clone SMP14, Chemicon®, from mouse
Sigma-Aldrich
Monoclonal Anti-β-Tubulin antibody produced in mouse, clone TUB 2.1, ascites fluid
Sigma-Aldrich
Etoposide, synthetic, 95.0-105.0%, powder
Sigma-Aldrich
MG-132, InSolution, ≥98%, 10 mM, reversible proteasome inhibitor