Skip to Content
MilliporeSigma
All Photos(1)

Documents

MAB3420A4

Sigma-Aldrich

Anti-Tau-1 Antibody, clone PC1C6, Alexa Fluor 488 Conjugate

clone PC1C6, from mouse, ALEXA FLUOR 488

Synonym(s):

Microtubule-associated protein tau, Neurofibrillary tangle protein, Paired helical filament-tau, PHF-tau

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

PC1C6, monoclonal

species reactivity

mouse, rat

species reactivity (predicted by homology)

bovine (immunogen homology)

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

Tau, a microtubulebinding protein which serves to stabilize microtubules in growing axons, is found to be hyperphosphorylated in paired helical filaments (PHF), the major fibrous component of neurofibrillary lesions associated with Alzheimer’s disease. Hyperphosphorylation of Tau is thought to be the critical event leading to the assembly of PHF. Six Tau protein isoforms have been identified, all of which are phosphorylated by glycogen synthase kinase 3 (GSK 3). Cellular and subcellular localization: In situ, anti-tau-1 has a stringent specificity for the axons of neurons. The antibody does not stain the cell bodies or dendrites of neurons, nor does it stain any other cell type (4). However, this in vivo intracellular specificity is not maintained in culture: anti-tau-1 stains the axon, cell bodies, and dendrites of rat hippocampal neurons grown in culture (5). The specificity of anti-tau-1 was originally thought to represent the restricted expression of tau to axons. Later studies revealed that this specificity is dependant on the state of phosphorylation. In dephosphorylated samples (samples treated with alkaline phosphatase) anti-tau-1 stains astrocytes, perineuronal glial cells, and the axons, cell bodies and dendrites of neurons, while in untreated samples, anti-tau-1 stains only axons (6). (The epitope recognized by anti-tau-1 is probably at or near a phosphorylated site.)

Specificity

This antibody binds to all known electrophoretic species of tau in human, rat, and bovine brain (one-dimensional SDS-PAGE).

Immunogen

Purified denatured bovine microtubule associated proteins.

Application

Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases
This Anti-Tau-1 Antibody, clone PC1C6, Alexa Fluor 488 Conjugate is validated for use in IC, IH for the detection of Tau-1.

Quality

Evaluated by Immunohistochemistry in adult mouse brain tissue.

Immunohistochemistry Analysis: A 1:100 dilution of this antibody detected Tau-1 in adult mouse brain tissue.

Target description

79 kDa calculated. Uniprot describes isoforms ranging from 33 to 81 kDa

Physical form

Protein A purified
Purified mouse monoclonal IgG1 conjugated to Biotin in PBS with 0.1% sodium azide and 15 mg/mL BSA.

Storage and Stability

Maintain refrigerated at 2-8 °C protected from light in undiluted aliquots for up to 6 months from date of receipt.

Analysis Note

Control
Adult mouse brain tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regulation of axon growth by the JIP1-AKT axis.
Dajas-Bailador, F; Bantounas, I; Jones, EV; Whitmarsh, AJ
Journal of Cell Science null
Neuroprotection and axon regeneration by novel low-molecular-weight compounds through the modification of DOCK3 conformation.
Namekata, et al.
Cell death discovery, 9, 166-166 (2023)
Dongdong Zhang et al.
Nature neuroscience, 27(2), 232-248 (2024-01-04)
Neurovascular coupling (NVC) is important for brain function and its dysfunction underlies many neuropathologies. Although cell-type specificity has been implicated in NVC, how active neural information is conveyed to the targeted arterioles in the brain remains poorly understood. Here, using

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service