Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB2239

Sigma-Aldrich

ANTI-TAU (MAPT) Antibody

mouse monoclonal, Tau 7

Synonym(s):

G protein beta1/gamma2 subunit-interacting factor, Neurofibrillary tangle protein, Paired helical filament-tau, microtubule-associated protein tau, microtubule-associated protein tau, isoform 4

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$610.00

$610.00


Please contact Customer Service for Availability
A recombinant, preservative-free antibody is available for your target. Try ZMS3420

Request a Bulk Order

Select a Size

Change View
100 μG
$610.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$610.00


Please contact Customer Service for Availability
A recombinant, preservative-free antibody is available for your target. Try ZMS3420

Request a Bulk Order

Product Name

Anti-Tau Antibody, clone Tau 7, clone Tau 7, from mouse

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

Tau 7, monoclonal

species reactivity

human, rat

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MAPT(4137)
rat ... Mapt(29477)

Related Categories

General description

Microtubule Associated Proteins, or MAPS, bind to the tubulin subunits of microtubule structures and regulate their functional stability. In the cell MAPs bind to monomer and multimerized tubulin. MAP binding to multimerized tubulin further stabilizes the formation of higher order microtubulin structures. MAP binding to microtubule structures is mediated through phosphorylation through Microtubule Affinity Regulated Kinase (MARK). Phosphorylation releases MAPs bound to microtubules, destabilizing the structure, driving it toward disassembly. There are predominately two MAP types, I, II. Type II MAP includes MAP2, MAP4, and tau and are found in nervous tissue. Six tau isoforms exist in brain tissue, and they are distinguished by their number of binding domains. Three isoforms have three binding domains and the other three have four binding domains. The binding domains are located in the carboxy-terminus of the protein and are positively-charged (allowing it to bind to the negatively-charged microtubule). The isoforms with four binding domains are better at stabilizing microtubules than those with three binding domains.

Specificity

Cat. # MAB2239 recognizes the C-terminus region of Tau.
Reactivity with other species has not been determined.

Application

This Anti-Tau Antibody, clone Tau 7 is validated for use in WB for the detection of Tau.

Quality

Western Blot:

Target description

50-68 kDa

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chiara Galimberti et al.
American journal of cancer research, 11(7), 3558-3574 (2021-08-07)
Glioblastoma multiforme (GBM) is the most malignant primary brain cancer. Despite aggressive treatments currently there is no cure for GBM. Many challenges should be considered for the development of new therapeutical agents for glioblastoma, including appropriate target selectivity and pharmacokinetics.
Takayuki Ohnishi et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(32), E4465-E4474 (2015-08-01)
Neurodegeneration correlates with Alzheimer's disease (AD) symptoms, but the molecular identities of pathogenic amyloid β-protein (Aβ) oligomers and their targets, leading to neurodegeneration, remain unclear. Amylospheroids (ASPD) are AD patient-derived 10- to 15-nm spherical Aβ oligomers that cause selective degeneration
Rik van der Kant et al.
Cell stem cell, 24(3), 363-375 (2019-01-29)
Genetic, epidemiologic, and biochemical evidence suggests that predisposition to Alzheimer's disease (AD) may arise from altered cholesterol metabolism, although the molecular pathways that may link cholesterol to AD phenotypes are only partially understood. Here, we perform a phenotypic screen for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service