Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB10201

Sigma-Aldrich

Anti-Hepatitis B Virus Surface Antigen Antibody, clone H25B10 | MAB10201

ascites fluid, clone H25B10, Chemicon®

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$613.00

$613.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$613.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$613.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

H25B10, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunoprecipitation (IP): suitable

isotype

IgG1

shipped in

dry ice

General description

Hepatitis B virus (HBV) is a small DNA virus belonging to the hepadnavirus family and is believed to the causative agent of type B hepatitis. The Hepatitis B surface antigen (HBsAg) is a protein antigen produced by HBV. This antigen is the earliest indicator of acute hepatitis B and frequently identifies infected people before symptoms appear.

Immunogen

Hepatitis B virus surface antigen

Application

Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral

Physical form

Ascites
Liquid ascites fluid containing no preservatives.

Storage and Stability

1 year at -20°C from date of shipment

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prevalence of anti HCV, HBsAg and HIV antibodies in high risk recipients of blood and blood products.
Chakrabarti, S, et al.
Indian Journal of Public Health, 50, 43-44 (2006)
Sang Hyun Nam et al.
Archives of pharmacal research, 29(11), 1042-1048 (2006-12-07)
Plasmid DNA vaccines encoding the hepatitis B virus (HBV) surface and hepatitis C virus (HCV) envelope antigens, respectively, were constructed, and attempt were made to find the possibility of a divalent vaccine against HBV and HCV. The expression of each

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service