K3388
Kappa Light Chains (Free) from human myeloma sera
purified immunoglobulin, ~1.0 mg/mL, ≥95% (SDS-PAGE), buffered aqueous solution
Synonym(s):
Free Kappa Light Chains, Kappa Light Chains
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
human
Quality Level
antibody form
purified immunoglobulin
Assay
≥95% (SDS-PAGE)
form
buffered aqueous solution
concentration
~1.0 mg/mL
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
General description
κ Light Chains (free) is obtained from human IgG κ myeloma sera. Fractionation, ion-exchange, and affinity chromatography procedures are used for purification.
Application
Purified κ light chains may be used as an immunoglobulin calibrator, reference antigen, blocking agent or coating protein in a variety of immunoassays including ELISA, dot-blot immunobinding, Western immunoblotting, immunodiffusion, immunoelectrophoresis, hemagglutination, and cell-binding assays.
Physical form
Solution in 0.01 M phosphate and 0.15 M sodium chloride, pH 7.2, containing 1.0 M glycine and 0.1% sodium azide.
Preparation Note
Purified from pooled IgG kappa myeloma sera.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service