Skip to Content
MilliporeSigma
All Photos(2)

Documents

HND3MAG-39K

Millipore

MILLIPLEX® Human Neurological Disorders Magnetic Bead Panel 3 - Neuroscience Multiplex Assay

The analytes available for this multiplex kit are: AGT, Contactin-1, Fetuin A, Kallikrein-6, OPN, sSOD1, sSOD2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

Quality Level

species reactivity

human

manufacturer/tradename

Milliplex®

assay range

accuracy: 69-92%
(Cerebrospinal fluid (CSF))

accuracy: 71-107%
(Serum)

standard curve range: 0.03-25 ng/mL
(SOD2)

standard curve range: 0.04-30 ng/mL
(Kallikrein-6)

standard curve range: 0.04-30 ng/mL
(SOD1)

standard curve range: 0.08-60 ng/mL
(Contactin-1)

standard curve range: 0.14-100 ng/mL
(Osteopontin (OPN))

standard curve range: 1.37-1,000 ng/mL
(AGT)

standard curve range: 4.12-3,000 ng/mL
(Fetuin A)

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex xMAP)

shipped in

wet ice

General description

Neurological disorders are diseases of the brain, spinal cord, and nerves. There are more than 600 diseases of the nervous system, such as Multiple sclerosis (MS), Alzheimer′s disease (AD), amyotrophic lateral sclerosis (ALS), Parkinson′s disease (PD), and stroke as well as frontotemporal dementia and many others. Identification of biomarkers for these neurological disorders allows scientists to identify early signs of neurological damage, track disease progression, and monitor efficacy of therapeutic treatment. The MILLIPLEX® 7-plex Human Neurological Disorders Bead Panel 3 is designed for the simultaneous measurement of multiple biomarkers in human cerebrospinal fluid (CSF), plasma, and serum using the Luminex® xMAP® instrument platform. The kit contains enough material to analyze 38 samples in duplicate, including the standard curve and two quality controls.

NOTE: For serum and plasma samples Fetuin A requires a higher (1:4000) dilution and therefore must be assayed alone as a singleplex. For serum or plasma samples all analytes, except Fetuin A, require a 1:50 dilution and may be assayed together as a multiplex.

The MILLIPLEX® Human Neurological Disorders Bead Panel 3 (Cat. No. HND3MAG-39K) is to be used for the simultaneous quantification of the following 7 analytes in any combination in CSF, serum or plasma samples: Angiotensinogen (AGT), Contactin-1, Fetuin A, Kallikrein-6, Osteopontin (OPN), Soluble Superoxide Dismutase 1 (SOD1), Soluble Superoxide Dismutase 2 (SOD2).

NOTE: For serum and plasma samples Fetuin A requires a higher (1:4000) dilution and therefore must be assayed alone as a singleplex. For serum or plasma samples all analytes, except Fetuin A, require a 1:50 dilution and may be assayed together as a multiplex.

Panel Type: Neuroscience

Specificity

Cross Reactivty
There was no or negligible cross-reactivity between the antibodies for an analyte and any of the other analytes in this panel.

Application

  • Analytes: Angiotensinogen (AGT), Contactin-1, Fetuin A, Kallikrein-6, Osteopontin (OPN), Soluble Superoxide Dismutase 1 (SOD1), Soluble Superoxide Dismutase 2 (SOD2)
  • Recommended Sample type: CSF, serum, and plasma
  • Recommended Sample dilution: 1:100 for CSF, 1:50 for serum and plasma for all analytes except Fetuin A, 1:4000 for serum and plasma for Fetuin A
  • NOTE: Fetuin A cannot be assayed along with other analytes in serum/plasma samples. Fetuin A should be diluted 1:4000 in the kit Assay Buffer and analyzed as a singleplex for serum and plasma samples
  • Assay Run Time: One day or Overnight
  • Research Category: Neuroscience

Features and Benefits

Design your multiplex kit by choosing available analytes within this panel.

Packaging

Everything you need in a single kit.

Storage and Stability

Recommended storage for kit components is 2 - 8°C.

Other Notes

Sensitivity: Refer to kit protocol for sensitivities of individual biomarkers.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Teresa Nestares et al.
Nutrients, 13(1) (2021-01-10)
The current study was designed to assess the influence of consumption of ultra-processed (UPF) on oxidative/antioxidant balance and evoked inflammatory signaling in young patients with celiac disease (CD). The study included 85 children. The celiac group (n = 53) included
Javier Diaz-Castro et al.
European journal of nutrition, 59(4), 1577-1584 (2019-05-31)
Celiac disease (CD) is a multifactorial, autoimmune, gluten-sensitive inflammatory disorder of the small intestine. Taking into account the pathogenesis of CD, a strict gluten-free diet (GFD) is the only treatment able to restore epithelium integrity and eliminate complications. The current

Related Content

Multiplex detection of Alzheimer's biomarkers with MILLIPLEX® Human Amyloid Beta and Tau Magnetic Bead Panel aids research.

Neuroscience multiplex assays measure biomarkers to understand neurodegenerative diseases, disorders, and neuropeptide signaling.

Find immunoassays to explore neuroimmunology and neuroinflammation cytokines and neurodegenerative disease biomarkers.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service