MilliporeSigma
  • Influence of Ultra-Processed Foods Consumption on Redox Status and Inflammatory Signaling in Young Celiac Patients.
Influence of Ultra-Processed Foods Consumption on Redox Status and Inflammatory Signaling in Young Celiac Patients.
Nutrients (2021-01-10)
Teresa Nestares, Rafael Martín-Masot, Marta Flor-Alemany, Antonela Bonavita, José Maldonado, Virginia A Aparicio
ABSTRACT
The current study was designed to assess the influence of consumption of ultra-processed (UPF) on oxidative/antioxidant balance and evoked inflammatory signaling in young patients with celiac disease (CD). The study included 85 children. The celiac group (n = 53) included children with CD with a long (>18 months, n = 17) or recent (<18 months, n = 36) adherence to a gluten-free diet (GFD). The control group (n = 32) included healthy children with a significantly higher consumption of UPF compared to the control group, both expressed as kcal/day (p = 0.043) and as percentage of daily energy intake (p = 0.023). Among children with CD, the group with the lowest consumption of UPF (below the 50% of daily energy intake) had a greater Mediterranean diet (MD) adherence and higher moderate physical activity levels. In addition, CD children with the lowest consumption of UPF had healthier redox (lower soluble superoxide dismutase-1 and 15-F2t-isoprostanes) and inflammatory profiles (lower macrophage inflammatory protein-1α) compared to the group with the highest consumption of UPF (all, p < 0.05) regardless of the time on a GFD. These findings highlight the importance of a correct monitoring of the GFD. An unbalanced GFD with high consumption of UPF and an unhealthy pattern with less physical activity and worse adherence to MD results in a worse inflammatory profile, which could act as a parallel pathway that could have important consequences on the pathophysiology of the disease.
Product Description
Millipore
MILLIPLEX® Human Neurological Disorders Magnetic Bead Panel 3 - Neuroscience Multiplex Assay, The analytes available for this multiplex kit are: AGT, Contactin-1, Fetuin A, Kallikrein-6, OPN, sSOD1, sSOD2